Merck
  • Abnormalities in Expression of Structural, Barrier and Differentiation Related Proteins, and Chondroitin Sulfate in Feline and Human Interstitial Cystitis.
Abnormalities in Expression of Structural, Barrier and Differentiation Related Proteins, and Chondroitin Sulfate in Feline and Human Interstitial Cystitis.
The Journal of urology (2015-02-01)
Paul J Hauser, Samuel B VanGordon, Jonathan Seavey, Troy M Sofinowski, Mohammad Ramadan, Shivon Abdullah, C A Tony Buffington, Robert E Hurst
ABSTRACT
We analyzed the urothelium of cats diagnosed with feline interstitial cystitis to determine whether abnormalities in protein expression patterns could be detected and whether the expression pattern was similar to that in patients with human interstitial cystitis/bladder pain syndrome. The proteins analyzed are involved in cell adhesion and barrier function, comprise the glycosaminoglycan layer or are differentiation markers. Formalin fixed biopsies from 8 cats with feline interstitial cystitis and from 7 healthy control cats were labeled by immunohistochemistry and scored with a modified version of a system previously used for human samples. Cluster analysis was performed to investigate relationships between markers and samples. Of the feline interstitial cystitis bladders 89% showed abnormal protein expression and chondroitin sulfate patterns while only 27% of normal tissues showed slight abnormalities. Abnormalities were found in most feline interstitial cystitis samples, including biglycan in 87.5%, chondroitin sulfate, decorin, E-cadherin and keratin-20 in 100%, uroplakin in 50% and ZO-1 in 87.5%. In feline interstitial cystitis bladders about 75% of chondroitin sulfate, biglycan and decorin samples demonstrated absent luminal staining or no staining. Cluster analysis revealed that feline interstitial cystitis and normal samples could be clearly separated into 2 groups, showing that the urothelium of cats with feline interstitial cystitis is altered from normal urothelium. Feline interstitial cystitis produces changes in luminal glycosaminoglycan and several proteins similar to that in patients, suggesting some commonality in mechanism. Results support the use of feline interstitial cystitis as a model of human interstitial cystitis.
Sigma-Aldrich
Hematoxylin
Sigma-Aldrich
Chondroitinase ABC from Proteus vulgaris, BSA free, lyophilized powder, specific activity 50-250 units/mg protein
Sigma-Aldrich
Hematoxylin, certified by the BSC
Sigma-Aldrich
Sodium acetate solution, BioUltra, Molecular Biology, ~3 M in H2O
Sigma-Aldrich
3-Amino-9-ethylcarbazole, ≥95% (HPLC), powder
Sigma-Aldrich
3-Amino-9-ethylcarbazole, tablet
Sigma-Aldrich
Sodium acetate, powder, BioReagent, suitable for electrophoresis, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Proteinase, bacterial, Type XXIV, 7.0-14.0 units/mg solid, lyophilized powder
Sigma-Aldrich
Sodium Acetate Anhydrous, >99%, FG
Sigma-Aldrich
Sodium acetate, 99.995% trace metals basis
Sigma-Aldrich
Sodium acetate, anhydrous, BioUltra, suitable for luminescence, Molecular Biology, ≥99.0% (NT)
Sigma-Aldrich
Sodium acetate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium acetate, anhydrous, Molecular Biology, ≥99%
Sigma-Aldrich
Sodium acetate, meets USP testing specifications, anhydrous