Merck
  • Aptamer-based microchip electrophoresis assays for amplification detection of carcinoembryonic antigen.
Aptamer-based microchip electrophoresis assays for amplification detection of carcinoembryonic antigen.
Clinica chimica acta; international journal of clinical chemistry (2015-09-08)
Li Pan, Jingjin Zhao, Yong Huang, Shulin Zhao, Yi-Ming Liu
ABSTRACT
Carcinoembryonic antigen (CEA) as one of the most widely used tumor markers is used in the clinical diagnosis of colorectal, pancreatic, gastric, and cervical carcinomas. We developed an aptamer-based microchip electrophoresis assay technique for assaying CEA in human serum for cancer diagnosis. The magnetic beads (MBs) are employed as carriers of double strand DNA that is formed by an aptamer of the target and a complementary DNA of the aptamer. After the aptamer in the MB-dsDNA conjugate binds with the target, the complementary DNA was released from the MB-dsDNA conjugate. The released complementary DNA hybridizes with a fluorescein amidite (FAM) labeled DNA, and forms a DNA duplex, which triggers the selective cleavage of FAM labeled DNA by nicking endonuclease Nb.BbvCI, and generating a FAM labeled DNA segment. The released complementary DNA hybridizes with another FAM labeled DNA, resulting in a continuous cleavage of FAM labeled DNA, and the generation of large numbers of FAM labeled DNA segments. In MCE laser induced fluorescence detection (LIF), the FAM labeled DNA segment is separated and detected. The linear range for CEA was 130 pg/ml-8.0 ng/ml with a correlation coefficient of 0.9916 and a detection limit of 68 pg/ml. The CEA concentration in the serum samples from healthy subjects was found to be in the range 1.3 ng/ml to 3.2 ng/ml. The CEA concentration in the samples from cancer patients was found to be >15 ng/ml. This method may become a useful tool for rapid analysis of CEA and other tumor markers in biomedical analysis and clinical diagnosis.
Sigma-Aldrich
1,2-Dichloroethane, ACS reagent, ≥99.0%
Sigma-Aldrich
1,2-Dichloroethane, anhydrous, 99.8%
Sigma-Aldrich
N-Hydroxysuccinimide, 98%
Sigma-Aldrich
MES solution, BioUltra, Molecular Biology, 0.5 M in H2O
Sigma-Aldrich
MES, low moisture content, ≥99% (titration)
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
Thiazole Orange, Dye content ~90 %
Sigma-Aldrich
Hydrochloric acid solution, for amino acid analysis, ~6 M in H2O
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in acetic acid
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrogen chloride solution, 4.0 M in dioxane
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, Molecular Biology
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrochloric acid, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrochloric acid, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
Fluorescein Phosphoramidite, configured for PerkinElmer, configured for Polygen
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in diethyl ether
Sigma-Aldrich
Hydrogen chloride solution, 2.0 M in diethyl ether