Merck
Sterol Composition in Infant Formulas and Estimated Intake.
Journal of agricultural and food chemistry (2015-08-06)
Lorena Claumarchirant, Esther Matencio, Luis Manuel Sanchez-Siles, Amparo Alegría, María Jesús Lagarda
ABSTRACT
Sterol contents in infant formulas (IFs) from the European market were determined, and their intakes by infants between 0 and 6 months were evaluated. Total animal sterols (mg/100 mL) ranged from 1.71 to 5.46, cholesterol being the main animal sterol (1.46-5.1). In general, cholesterol and desmosterol were lower than the human milk (HM) values indicated by other authors. Total plant sterol (mg/100 mL) ranged from 3.1 to 5.0. β-Sitosterol, the most abundant phytosterol, ranged from 1.82 to 3.01, followed by campesterol (0.72-1.15), stigmasterol (0.27-0.53), and brassicasterol (0.14-0.28). Cholesterol intake (mg/day) ranged from 9 to 51 and plant sterol intake (mg/day) from 19 to 50. The sterol profile of IFs is highly dependent on the type and quantity of fats used in their formula. The use of bovine milk fat and milk fat globule membrane in the IFs can approximate the profile of animal sterols to those found in HM, though cholesterol intakes in breastfed infants are still higher than in formula-fed infants.
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Acetone, ≥99%, FCC, FG
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Butylated hydroxytoluene, ≥99%, FCC, FG
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Hexamethyldisiloxane, NMR grade, ≥99.5%
Sigma-Aldrich
Potassium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Chlorotrimethylsilane, Wacker Chemie AG, ≥99.0% (GC)
Sigma-Aldrich
Potassium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Chlorotrimethylsilane, ≥98.0% (GC)
Sigma-Aldrich
Sodium sulfate, BioUltra, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Diethyl ether
Sigma-Aldrich
Potassium chloride, ≥99.99% trace metals basis
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Potassium chloride, 99.999% trace metals basis
Sigma-Aldrich
Hexamethyldisiloxane, ≥98%
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Potassium chloride, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture