Merck
  • Effects of Eicosapentaenoic Acid on the Cytoprotection Through Nrf2-Mediated Heme Oxygenase-1 in Human Endothelial Cells.
Effects of Eicosapentaenoic Acid on the Cytoprotection Through Nrf2-Mediated Heme Oxygenase-1 in Human Endothelial Cells.
Journal of cardiovascular pharmacology (2015-03-31)
Seung Eun Lee, Gun-Dong Kim, Hana Yang, Gun Woo Son, Hye Rim Park, Jeong-Je Cho, Hyun-Jong Ahn, Cheung-Seog Park, Yong Seek Park
ABSTRACT
Consumption of omega-3 polyunsaturated fatty acid, particularly eicosapentaenoic acid (EPA), is associated with a significant reduction in the risk of developing cardiovascular disease. The aim of this study was to investigate whether heme oxygenase-1 (HO-1) induction contributes to the cytoprotective effects of EPA in endothelial cells threatened with oxidative damage. In this study, we investigated the effect of EPA on the induction of HO-1 by NF-E2-related factor 2 (Nrf2) in human umbilical vein endothelial cells. In cells treated with low concentrations of EPA (10-25 μM), HO-1 expression increased in a time- and concentration-dependent manner. Additionally, EPA treatment increased Nrf2 nuclear translocation and antioxidant response element activity, leading to the upregulation of HO-1 expression. Furthermore, treatment with EPA reduced hydrogen peroxide (H(2)O(2))-induced cell death. The reduction in cell death was reversed by treatment with zinc protoporphyrin, an inhibitor of HO-1, indicating that HO-1 contributed to the protective effect of EPA. These data suggest that EPA protects against H(2)O(2)-induced oxidative stress in endothelial cells by activating Nrf2 and inducting HO-1 expression.
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
Tetramethylrhodamine ethyl ester perchlorate, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)