Merck
  • Evidence that the inhibition sites of the neurotoxic amine 1-methyl-4-phenylpyridinium (MPP+) and of the respiratory chain inhibitor piericidin A are the same.
Evidence that the inhibition sites of the neurotoxic amine 1-methyl-4-phenylpyridinium (MPP+) and of the respiratory chain inhibitor piericidin A are the same.
The Biochemical journal (1991-01-15)
R R Ramsay, M J Krueger, S K Youngster, T P Singer
ABSTRACT
1-Methyl-4-phenylpyridinium (MPP+), the neurotoxic bioactivation product of 1-methyl-4-phenyl-1,2,3,6-tetrahydropyridine (MPTP), interrupts mitochondrial electron transfer at the NADH dehydrogenase-ubiquinone junction, as do the respiratory chain inhibitors rotenone, piericidin A and barbiturates. Proof that these classical respiratory chain inhibitors and MPP+ react at the same site in the complex NADH dehydrogenase molecule has been difficult to obtain because none of these compounds bind covalently to the target. The 4'-alkyl derivatives of MPP+ inhibit NADH oxidation in submitochondrial particles at much lower concentrations than does MPP+ itself, but still dissociate on washing the membrane preparations, with consequent re-activation of the enzyme. The MPP+ analogues with short alkyl chains prevent the binding of 14C-labelled piericidin A to the membrane and thus must act at the same site, but analogues with alkyl chains longer than heptyl do not prevent binding of [14C]piericidin.