Merck
  • Neonatal vaccination with bacillus Calmette-Guérin and hepatitis B vaccines modulates hippocampal synaptic plasticity in rats.
Neonatal vaccination with bacillus Calmette-Guérin and hepatitis B vaccines modulates hippocampal synaptic plasticity in rats.
Journal of neuroimmunology (2015-11-05)
Qingqing Li, Fangfang Qi, Junhua Yang, Luwen Zhang, Huaiyu Gu, Juntao Zou, Qunfang Yuan, Zhibin Yao
ABSTRACT
Immune activation can exert multiple effects on synaptic transmission. Our study demonstrates the influence of neonatal vaccination on hippocampal synaptic plasticity in rats under normal physiological conditions. The results revealed that neonatal BCG vaccination enhanced synaptic plasticity. In contrast, HBV hampered it. Furthermore, we found that the cytokine balance shifted in favour of the T helper type 1/T helper type 2 immune response in BCG/HBV-vaccinated rats in the periphery. The peripheral IFN-γ:IL-4 ratio was positively correlated with BDNF and IGF-1 in the hippocampus. BCG raised IFN-γ, IL-4, BDNF and IGF-1 and reduced IL-1β, IL-6, and TNF-α in the hippocampus, whereas, HBV triggered the opposite effects.
Sigma-Aldrich
Sodium azide, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Sodium azide, BioXtra
Sigma-Aldrich
Sodium azide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium azide, purum p.a., ≥99.0% (T)
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
Calcium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Calcium chloride
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
Calcium chloride, powder, 99.99% trace metals basis