  • Search Results
  • WNT5A promotes stemness characteristics in nasopharyngeal carcinoma cells leading to metastasis and tumorigenesis.
WNT5A promotes stemness characteristics in nasopharyngeal carcinoma cells leading to metastasis and tumorigenesis.
Oncotarget (2015-04-01)
Li Qin, Yan-Tao Yin, Fang-Jing Zheng, Li-Xia Peng, Chang-Fu Yang, Ying-Na Bao, Ying-Ying Liang, Xin-Jian Li, Yan-Qun Xiang, Rui Sun, An-Hua Li, Ru-Hai Zou, Xiao-Qing Pei, Bi-Jun Huang, Tie-Bang Kang, Duan-Fang Liao, Yi-Xin Zeng, Bart O Williams, Chao-Nan Qian
ABSTRACT
Nasopharyngeal carcinoma (NPC) has the highest metastasis rate among head and neck cancers with unclear mechanism. WNT5A belongs to the WNT family of cysteine-rich secreted glycoproteins. Our previous high-throughput gene expression profiling revealed that WNT5A was up-regulated in highly metastatic cells. In the present study, we first confirmed the elevated expression of WNT5A in metastatic NPC tissues at both the mRNA and protein levels. We then found that WNT5A promoted epithelial-mesenchymal transition (EMT) in NPC cells, induced the accumulation of CD24-CD44+ cells and side population, which are believed to be cancer stem cell characteristics. Moreover, WNT5A promoted the migration and invasion of NPC cells in vitro, while in vivo treatment with recombinant WNT5A promoted lung metastasis. Knocking down WNT5A diminished NPC tumorigenesis in vivo. When elevated expression of WNT5A coincided with the elevated expression of vimentin in the primary NPC, the patients had a poorer prognosis. Among major signaling pathways, protein kinase C (PKC) signaling was activated by WNT5A in NPC cells. A positive feedback loop between WNT5A and phospho-PKC to promote EMT was also revealed. Taken together, these data suggest that WNT5A is an important molecule in promoting stem cell characteristics in NPC, leading to tumorigenesis and metastasis.
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
MISSION® pLKO.1-puro Non-Mammalian shRNA Control Plasmid DNA, Targets no known mammalian genes
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
SAFC
L-Glutamine
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, synthetic, ≥98.0% (TLC)
Sigma-Aldrich
PMA, for use in molecular biology applications, ≥99% (HPLC)
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, ≥99% (TLC), film or powder