Merck
  • Apoptosis induced by weisiensin B isolated from Rabdosia weisiensis C.Y. Wu in K562.
Apoptosis induced by weisiensin B isolated from Rabdosia weisiensis C.Y. Wu in K562.
Die Pharmazie (2015-05-28)
Guo-An Liu, Jin-Chun Chang, Xiao-Lu Feng, Lan Ding
ABSTRACT
The ent-kaurane diterpenoid weisinensis B shows significant cytotoxicity to human chronic myeloid leukemia K562 cells. It inhibits cell growth at low concentration and kills cells at high concentration. The compound induced cell apoptosis and necrosis mainly associated with G2/M phase cell cycle arrest and the ROS generation is the early event in weisiensin B induced cell apoptosis.
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Sulforhodamine B, Dye content 75 %
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
Sulforhodamine B sodium salt, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Sulforhodamine B sodium salt, Technical grade