Merck
  • In vitro cytotoxicity evaluation of HDAC inhibitor Apicidin in pancreatic carcinoma cells subsequent time and dose dependent treatment.
In vitro cytotoxicity evaluation of HDAC inhibitor Apicidin in pancreatic carcinoma cells subsequent time and dose dependent treatment.
Toxicology letters (2015-04-29)
Monika Bauden, Helena Tassidis, Daniel Ansari
ABSTRACT
Apicidin is a potent histone deacetylase inhibitor (HDACI) that selectively binds to histone deacetylases (HDACs) class I and interferes with the deacetylation process, which results in modification of acetylation level of cellular proteins. The aim of the study was to investigate the potential time and dose dependent cytotoxicity of the test compound, Apicidin, in pancreatic cancer cells Capan-1 and Panc-1 as well as estimate maximal tolerable dose (MTD) of the test agent and determine EC50 using four complementary colorimetric cytotoxicity or viability assays. The cells were treated with increasing concentrations of Apicidin (0-5000nM) for 2, 4 and 6h (short term exposure) or 24, 48 and 72h (long term exposure) before conducting cytotoxic analyses with lactate dehydrogenase assay or viability analyses with sulforhodamine B (SRB), methyl tetrazolium (MTT) and crystal violet (CV) assays. In order to investigate whether Apicidin irreversibly affects the cells already during the short term exposure, the medium containing Apicidin was removed and replaced with fresh culturing medium after 6h of treatment. The cells were then incubated for additional 24, 48 or 72h before carrying out the analysis. The results obtained from cytotoxicity and viability assays indicated, that Apicidin was well tolerated by both cell lines at concentrations below 100nM at any given time point and at all applied concentrations during the short term (6h or less) treatment. Continuous prolonged term exposures (48h or greater) of the cells to Apicidin with concentration exceeding 100nM resulted in significantly increasing cytotoxicity and sustained significant loss of cell viability. Moreover, long term exposure of pancreatic cancer cells Capan-1 and Panc-1 to Apicidin concentrations exceeding 100nM showed an initial anti-proliferative effect before cytotoxicity onset. In summary, MTD was exposure time dependent and estimated to 100nM for long term treatment and to at least 5000nM for treatment not greater than 6h. EC50 concentration of Apicidin was established after long term treatment, however with some variation when comparing the different assays and cell lines. Results from this study may encourage reinvestigating the capacity of potent HDACI Apicidin as an attractive agent for interfering with the deacetylation process catalyzed by HDACs for potential pancreatic cancer intervention.
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Tromethamine, meets USP testing specifications
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Trizma® base, BioPerformance Certified, meets EP, USP testing specifications, suitable for cell culture, ≥99.9% (titration)
Sigma-Aldrich
Trizma® base, pH 10.5-12.0 (1 M in H2O), ≥99.9% (titration)
Sigma-Aldrich
Tris(hydroxymethyl)aminomethane, ACS reagent, ≥99.8%
Sigma-Aldrich
Sodium trichloroacetate, 97%
Sigma-Aldrich
Trizma® base, BioUltra, Molecular Biology, ≥99.8% (T)
Sigma-Aldrich
Trizma® base, ≥99.0% (T)
Sigma-Aldrich
Trizma® base, anhydrous, free-flowing, Redi-Dri, ≥99.9%
Sigma-Aldrich
Trizma® base, ≥99.9% (titration), crystalline
Sigma-Aldrich
Trizma® base, Primary Standard and Buffer, ≥99.9% (titration), crystalline
Sigma-Aldrich
Trizma® base, 99.7% (T), puriss. p.a.
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Apicidin, ≥98% (HPLC), from microbial
SAFC
Tromethamine
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Sigma 7-9®, ≥99% (titration), powder or crystals
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®