Merck
  • Determining canine skin concentrations of terbinafine to guide the treatment of Malassezia dermatitis.
Determining canine skin concentrations of terbinafine to guide the treatment of Malassezia dermatitis.
Veterinary dermatology (2015-08-20)
Jacqueline R Gimmler, Amelia G White, Robert A Kennis, Crisanta Cruz-Espindola, Dawn M Boothe
ABSTRACT
Terbinafine (TBF) is known to concentrate and persist in human skin. Its use is increasing in veterinary medicine, but there are limited data concerning its tissue concentration and efficacy in dogs. (i) Describe TBF accumulation in canine skin; (ii) Integrate pharmacokinetic data with historical minimum inhibitory concentration (MIC) results for Malassezia pachydermatis to verify the currently used dosage of TBF for the treatment of Malassezia dermatitis. Ten healthy, client-owned dogs. Dogs were given TBF (generic preparation, 250 mg tablets) 30 mg/kg per os (p.o.) once daily for 21 days. Serum, sebum and stratum corneum (SC) samples were collected on days 1, 5, 7, 11, 14, 21, 28 and 35. High-pressure liquid chromatography was used to determine drug concentrations in samples. Relevant (mean ± standard deviation) parameters for TBF in serum, paw SC, thorax SC and sebum, respectively, were: maximum concentration (Cmax , μg/mL) 23.59 ± 10.41, 0.31 ± 0.26, 0.30 ± 0.32 and 0.48 ± 0.25; half-life (t1/2 , d) 4.49 ± 2.24, 6.34 ± 5.33, 4.64 ± 3.27 and 5.12 ± 3.33; time to maximum concentration (Tmax , d) 10.40 ± 6.98, 13.20 ± 5.16, 11.90 ± 8.62 and 10.60 ± 3.69. These results suggest that TBF does not achieve high concentrations in canine SC or sebum compared to serum. The mean Cmax of all skin tissues (paw SC, thorax SC and sebum) barely exceeded the reported Malassezia MIC90, of 0.25 μg/mL, which indicates that doses higher than 30 mg/kg p.o. once daily may be necessary.
Sigma-Aldrich
Phosphoric acid-16O4 solution, 70 wt. % in D2O, 99.9 atom % 16O
Sigma-Aldrich
Phosphoric acid, BioReagent, suitable for insect cell culture, 85%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Phosphoric acid, ACS reagent, ≥85 wt. % in H2O
Sigma-Aldrich
Ammonium hydroxide solution, ACS reagent, 28.0-30.0% NH3 basis
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Phosphoric acid, ≥85 wt. % in H2O, ≥99.999% trace metals basis
Sigma-Aldrich
Phosphoric acid, 85 wt. % in H2O, 99.99% trace metals basis
Sigma-Aldrich
Phosphoric acid, crystalline, ≥99.999% trace metals basis
Sigma-Aldrich
Phosphoric acid, BioUltra, ≥85% (T)
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Phosphoric acid, ACS reagent, ≥85 wt. % in H2O
Sigma-Aldrich
Ammonium hydroxide solution, 28% NH3 in H2O, ≥99.99% trace metals basis
Sigma-Aldrich
Phosphoric acid solution, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 4.2 mm × 8 in. , WGS-5BL Coaxial NMR tube
Sigma-Aldrich
Phosphoric acid solution, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Phosphoric acid solution, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Triethylamine, for amino acid analysis, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, ampule, ≥99.5% (GC)
Sigma-Aldrich
Phosphoric acid solution, 85 wt. % in H2O, FCC, FG
Sigma-Aldrich
Triethylamine, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, ≥99%
Sigma-Aldrich
Triethylamine, ≥99.5%