Merck
  • Determination of N,N-dimethyldodecylamine and N,N-dimethyloctadecylamine in river and sea water using liquid chromatography tandem mass spectrometry.
Determination of N,N-dimethyldodecylamine and N,N-dimethyloctadecylamine in river and sea water using liquid chromatography tandem mass spectrometry.
Bulletin of environmental contamination and toxicology (2015-01-22)
Hiroko Tsukatani, Kazuhiro Tobiishi
ABSTRACT
We developed a selective method for determining N,N-dimethyldodecylamine (DMDA) and N,N-dimethyloctadecylamine (DMOA) concentrations in river and sea water samples using liquid chromatography/tandem mass spectrometry (LC/MS/MS). DMDA and DMOA are suspected to be toxic, and DMDA is categorized as a "Class I Designated Chemical Substance" under the "Act on Confirmation, etc. of Release Amounts of Specific Chemical Substances in the Environment and Promotion of Improvements to the Management Thereof" in Japan. The analytes are extracted from a water sample using solid phase extraction and the extract is evaporated and dissolved in 1 mL methanol for LC/MS/MS analysis. We analyzed DMDA and DMOA in real water samples and found that the analyte peaks were resolved effectively. The method detection limits for DMDA and DMOA were 4.7 and 0.80 ng/L, respectively.
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Phthalic acid, reagent grade, 98%
Sigma-Aldrich
Phthalic acid, puriss. p.a., ≥99.5% (T)
Sigma-Aldrich
N,N-Dimethyloctylamine, 95%
Sigma-Aldrich
Phthalic acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Sodium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Hydrochloric acid solution, for amino acid analysis, ~6 M in H2O
Sigma-Aldrich
Sodium sulfate, BioXtra, ≥99.0%
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, Molecular Biology
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium sulfate, ≥99.0%, suitable for plant cell culture
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Sodium sulfate, BioUltra, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Acetone, ≥99%, FCC, FG
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG