Merck
  • Optimization and Validation of an ETAAS Method for the Determination of Nickel in Postmortem Material.
Optimization and Validation of an ETAAS Method for the Determination of Nickel in Postmortem Material.
Journal of analytical toxicology (2015-04-15)
Danuta Dudek-Adamska, Teresa Lech, Paweł Kościelniak
ABSTRACT
In this article, optimization and validation of a procedure for the determination of total nickel in wet digested samples of human body tissues (internal organs) for forensic toxicological purposes are presented. Four experimental setups of the electrothermal atomic absorption spectrometry (ETAAS) using a Solaar MQZe (Thermo Electron Co.) were compared, using the following (i) no modifier, (ii) magnesium nitrate, (iii) palladium nitrate and (iv) magnesium nitrate and ammonium dihydrogen phosphate mixture as chemical modifiers. It was ascertained that the ETAAS without any modifier with 1,300/2,400°C as the pyrolysis and atomization temperatures, respectively, can be used to determine total nickel at reference levels in biological materials as well as its levels found in chronic or acute poisonings. The method developed was validated, obtaining a linear range of calibration from 0.76 to 15.0 μg/L, limit of detection at 0.23 µg/L, limit of quantification at 0.76 µg/L, precision (as relative standard deviation) up to 10% and accuracy of 97.1% for the analysis of certified material (SRM 1577c Bovine Liver) and within a range from 99.2 to 109.9% for the recovery of fortified liver samples.
Sigma-Aldrich
Ammonium phosphate monobasic, suitable for plant cell culture
Sigma-Aldrich
Ammonium dihydrogenphosphate, 99.999% trace metals basis
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer