Merck
  • Faceted phospholipid vesicles tailored for the delivery of Santolina insularis essential oil to the skin.
Faceted phospholipid vesicles tailored for the delivery of Santolina insularis essential oil to the skin.
Colloids and surfaces. B, Biointerfaces (2015-06-10)
Ines Castangia, Maria Letizia Manca, Carla Caddeo, Andrea Maxia, Sergio Murgia, Ramon Pons, Davide Demurtas, Daniel Pando, Danilo Falconieri, José E Peris, Anna Maria Fadda, Maria Manconi
ABSTRACT
The aim of this work was to formulate Santolina insularis essential oil-loaded nanocarriers, namely Penetration Enhancer containing Vesicles (PEVs), evaluate the physico-chemical features and stability, and gain insights into their ability to deliver the oil to the skin. S. insularis essential oil was obtained by steam distillation, and was predominantly composed of terpenes, the most abundant being β-phellandrene (22.6%), myrcene (11.4%) and curcumenes (12.1%). Vesicles were prepared using phosphatidylcholine, and ethylene or propylene glycol were added to the water phase (10% (v/v)) to improve vesicle performances as delivery systems. Vesicles were deeply characterized by light scattering, cryogenic transmission electron microscopy and small/wide-angle X-ray scattering, the results showing polyhedral, faceted, unilamellar vesicles of ∼115 nm in size. The presence of the glycols improved vesicle stability under accelerated ageing conditions, without changes in size or migration phenomena (e.g. sedimentation and creaming). Confocal laser scanning microscopy images of pig skin treated with S. insularis formulations displayed a penetration ability of PEVs greater than that of control liposomes. Moreover, all formulations showed a marked in vitro biocompatibility in human keratinocytes. These findings suggest that the nanoformulation may be of value in enhancing the delivery of S. insularis essential oil to the skin, where it can exert its biological activities.
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications
Sigma-Aldrich
Ethylene glycol, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Propylene glycol, ≥99.5% (GC), FCC, FG
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Propylene Glycol, meets USP testing specifications
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
Ethylene glycol, anhydrous, 99.8%
Supelco
Ethylene glycol solution, suitable for NMR (reference standard), 80% in DMSO-d6 (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
1,2-Propanediol, ReagentPlus®, 99%
Sigma-Aldrich
1,2-Propanediol, puriss. p.a., ACS reagent, ≥99.5% (GC)
Sigma-Aldrich
1,3-Propanediol, 98%
Sigma-Aldrich
1,2-Propanediol, meets analytical specification of Ph. Eur., BP, USP, ≥99.5%
Sigma-Aldrich
Ethylene glycol 5 M solution
Sigma-Aldrich
1,2-Propanediol, ACS reagent, ≥99.5%