Merck
  • Effect of novel restoration techniques on the fracture resistance of teeth treated endodontically: An in vitro study.
Effect of novel restoration techniques on the fracture resistance of teeth treated endodontically: An in vitro study.
Dental materials journal (2015-10-07)
Hande Kemaloglu, Mehmet Emin Kaval, Murat Turkun, Seniha Micoogullari Kurt
ABSTRACT
The aim was to evaluate the effects of fiber-reinforced composite restorations and a bulk-fill resin composite on the fracture strength of mandibular premolars treated endodontically. Standard mesio-occluso-distal (MOD) cavities were prepared in 48 mandibular premolars. Following root canal treatment, teeth were assigned to four groups: Group 1, nano-hybrid resin composite; Group 2, polyethylene woven fiber plus nano-hybrid resin composite; Group 3, short fiber-reinforced resin composite plus nano-hybrid resin composite; and Group 4, bulk-fill resin composite plus nano-hybrid resin composite. Then, the teeth were subjected to the fracture toughness test. The data were analyzed statistically using one-way ANOVA, followed by Tukey's post-hoc test. The fiber-reinforced groups had better results than the nano-hybrid and bulk-fill composites (p<0.05), while the bulk-fill and nano-hybrid composite restorations gave similar results (p>0.05). Fiber-reinforcement improved the fracture strength of teeth with large MOD cavities treated endodontically. Bulk-fill composites can be used reliably as well as nano-hybrid composites.
Sigma-Aldrich
Phosphoric acid solution, 85 wt. % in H2O, FCC, FG
Sigma-Aldrich
Phosphoric acid solution, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 4.2 mm × 8 in. , WGS-5BL Coaxial NMR tube
Sigma-Aldrich
Phosphoric acid solution, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Phosphoric acid solution, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Phosphoric acid, 85 wt. % in H2O, 99.99% trace metals basis
Sigma-Aldrich
Phosphoric acid, crystalline, ≥99.999% trace metals basis
Sigma-Aldrich
Phosphoric acid-16O4 solution, 70 wt. % in D2O, 99.9 atom % 16O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Phosphoric acid, BioReagent, suitable for insect cell culture, 85%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Phosphoric acid, BioUltra, ≥85% (T)
Sigma-Aldrich
Phosphoric acid, ≥85 wt. % in H2O, ≥99.999% trace metals basis
Sigma-Aldrich
Phosphoric acid, ACS reagent, ≥85 wt. % in H2O
Sigma-Aldrich
Phosphoric acid, ACS reagent, ≥85 wt. % in H2O