Merck
  • Effects of Endocrine Disruptor Compounds, Alone or in Combination, on Human Macrophage-Like THP-1 Cell Response.
Effects of Endocrine Disruptor Compounds, Alone or in Combination, on Human Macrophage-Like THP-1 Cell Response.
PloS one (2015-07-03)
N Couleau, J Falla, A Beillerot, E Battaglia, M D'Innocenzo, S Plançon, P Laval-Gilly, A Bennasroune
ABSTRACT
The aim of the present study was to evaluate the immunological effects on human macrophages of four endocrine disruptor compounds (EDCs) using the differentiated human THP-1 cell line as a model. We studied first the effects of these EDCs, including Bisphenol A (BPA), di-ethylhexyl-phthalate (DEHP), dibutyl phthalate (DBP) and 4-tert-octylphenol (4-OP), either alone or in combination, on cytokine secretion, and phagocytosis. We then determined whether or not these effects were mediated by estrogen receptors via MAPK pathways. It was found that all four EDCs studied reduced strongly the phagocytosis of the differentiated THP-1 cells and that several of these EDCs disturbed also TNF-α, IL-1 β and IL-8 cytokine secretions. Furthermore, relative to control treatment, decreased ERK 1/2 phosphorylation was always associated with EDCs treatments-either alone or in certain combinations (at 0.1 μM for each condition). Lastly, as treatments by an estrogen receptor antagonist suppressed the negative effects on ERK 1/2 phosphorylation observed in cells treated either alone with BPA, DEHP, 4-OP or with the combined treatment of BPA and DEHP, we suggested that estrogen receptor-dependent pathway is involved in mediating the effects of EDCs on human immune system. Altogether, these results advocate that EDCs can disturb human immune response at very low concentrations.
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, ≥99% (TLC), film or powder
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
PMA, for use in molecular biology applications, ≥99% (HPLC), Molecular Biology
Sigma-Aldrich
4-tert-Octylphenol, 97%
Sigma-Aldrich
Sodium Fluoride Solution
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Dibutyl phthalate, 99%
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, synthetic, ≥98.0% (TLC)
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
SAFC
L-Glutamine
Sigma-Aldrich
Sodium fluoride, BioXtra, ≥99%
Sigma-Aldrich
Sodium fluoride, BioReagent, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)
Sigma-Aldrich
DBP, 98% (HPLC)
Supelco
Bisphenol A, ≥99%
Sigma-Aldrich
Bisphenol A, 97%
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Glycerol, Molecular Biology, ≥99.0%
Sigma-Aldrich
Glycerol, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerol, ≥99.5%