Merck
  • Patchable, flexible heat-sensing hybrid ionic gate nanochannel modified with a wax-composite.
Patchable, flexible heat-sensing hybrid ionic gate nanochannel modified with a wax-composite.
Nanoscale (2015-07-02)
Kyoung-Yong Chun, Wook Choi, Sung-Cheoul Roh, Chang-Soo Han
ABSTRACT
Heat-driven ionic gate nanochannels have been recently demonstrated, which exploit temperature-responsive polymer brushes based on wettability. These heat-sensing artificial nanochannels operate in a broad temperature-response boundary and fixed liquid cell environment, thereby experiencing limited system operation in the flat and solid state. Here we have developed a patchable and flexible heat-sensing artificial ionic gate nanochannel, which can operate in the range of the human body temperature. A wax-elastic copolymer, coated onto a commercial nanopore membrane by a controlled-vacuum filtration method, was used for the construction of temperature-responsive nanopores. The robust and flexible nanochannel heat sensor, which is combined with an agarose gel electrolyte, can sustain reversible thermo-responsive ionic gating based on the volumetric work of the wax-composite layers in a selective temperature range. The ionic current is also effectively distinguished in the patchable bandage-type nanochannel for human heat-sensing.
Sigma-Aldrich
Potassium chloride, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Potassium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Potassium chloride, ≥99.99% trace metals basis
Sigma-Aldrich
Potassium chloride, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Potassium chloride, 99.999% trace metals basis
Sigma-Aldrich
Potassium chloride, Molecular Biology, ≥99.0%
Sigma-Aldrich
Potassium chloride solution, 0.075 M, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis