Merck
  • Glucose encapsulating liposome for signal amplification for quantitative detection of biomarkers with glucometer readout.
Glucose encapsulating liposome for signal amplification for quantitative detection of biomarkers with glucometer readout.
Biosensors & bioelectronics (2015-05-26)
Yuting Zhao, Dan Du, Yuehe Lin
ABSTRACT
A new technology was developed to quantitatively detect a broad range of disease biomarkers and proven to be portable, economical, and conveniently accessible. Measurements were performed based on releasing encapsulated glucose from antibody-tagged liposomes and subsequently detecting the released glucose using a commercial personal glucose meter (GM). The innovative aspect of this approach lies in the quantification of target biomarkers through the detection of glucose, thus expanding the applicability of the GM by broadening the range of target biomarkers instead of detecting only one analyte, glucose. Because of the bilayer membrane of liposomes, which can accommodate tens of thousands of glucose molecules, the sensitivity was greatly enhanced by using glucose encapsulating liposomes as a signal output and an amplifier. Here, the model analyte, protein 53 phosphorylated on Serine 15 (phospho-p53(15)), was captured by primary antibodies bound on magnetic Fe3O4 nanoparticles and then recognized by reporting antibodies conjugated to glucose encapsulating liposomes. Finally, the target phospho-p53(15) was detected by lysing the bound liposomes to release the encapsulated glucose (4 × 10(5) glucose molecules per liposome), which is detected with the GM. This approach was demonstrated to be a universal technology that can be easily produced to quantify a wide variety of biomarkers in medical diagnostics, food safety, public health, and environmental monitoring. In the near future, it is expected that these sensors, in combination with a portable GM, can be used in many fields such as physicians' laboratories, hospitals and the common household.
Sigma-Aldrich
MES solution, BioUltra, Molecular Biology, 0.5 M in H2O
Sigma-Aldrich
L-Lysine monohydrochloride, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
MES, low moisture content, ≥99% (titration)
Sigma-Aldrich
L-Lysine monohydrochloride, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Lysine monohydrochloride, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
N-(3-Dimethylaminopropyl)-N′-ethylcarbodiimide hydrochloride, ≥99.0% (AT)
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
N-Hydroxysuccinimide, 98%
Sigma-Aldrich
N-(3-Dimethylaminopropyl)-N′-ethylcarbodiimide hydrochloride, commercial grade, powder
Sigma-Aldrich
N-(3-Dimethylaminopropyl)-N′-ethylcarbodiimide hydrochloride, BioXtra
Sigma-Aldrich
N-(3-Dimethylaminopropyl)-N′-ethylcarbodiimide hydrochloride, crystalline
Sigma-Aldrich
1,2-Dichloroethane, anhydrous, 99.8%
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Sigma-Aldrich
(±)-2-(p-Methoxyphenoxy)propionic acid, ≥98%
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications