Merck
  • Organochlorine pesticides, lead, and mercury in northern bobwhite (Colinus virginianus) and scaled quail (Callipepla squamata) from the Rolling Plains ecoregion of Texas and Oklahoma.
Organochlorine pesticides, lead, and mercury in northern bobwhite (Colinus virginianus) and scaled quail (Callipepla squamata) from the Rolling Plains ecoregion of Texas and Oklahoma.
Environmental toxicology and chemistry (2015-02-11)
Catherine E Baxter, Sara Pappas, Michael T Abel, Ronald J Kendall
ABSTRACT
Northern bobwhite (Colinus virginianus) and scaled quail (Callipepla squamata) from the Rolling Plains ecoregion in Texas and Oklahoma were evaluated for organochlorine pesticides, Pb, and Hg. Of all organochlorine pesticides analyzed, only p,p'-dichlorodiphenyldichloroethylene (p,p'-DDE) and p,p'-dichlorodiphenyldichloroethane (p,p'-DDD) were found in a few composite liver samples. Similarly, a small fraction of tissue samples had detectable levels of Hg (liver and breast) or Pb exceeding background concentrations (femur). Lead concentrations in a few individuals fell within the range associated with moderate toxicity.
Sigma-Aldrich
Poly(tetrafluoroethylene), powder (free-flowing), ≤12 μm particle size
Sigma-Aldrich
Poly(tetrafluoroethylene), powder, 35 μm particle size
Sigma-Aldrich
Poly(tetrafluoroethylene), powder, 200 μm particle size
Sigma-Aldrich
Poly(tetrafluoroethylene), powder, >40 μm particle size
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Poly(tetrafluoroethylene), beads
Sigma-Aldrich
Poly(tetrafluoroethylene), powder, ≥350 μm particle size
Sigma-Aldrich
Poly(tetrafluoroethylene), powder (free-flowing), 1 μm particle size
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis