Merck
  • Inhibitory mechanisms of celastrol on human liver cytochrome P450 1A2, 2C19, 2D6, 2E1 and 3A4.
Inhibitory mechanisms of celastrol on human liver cytochrome P450 1A2, 2C19, 2D6, 2E1 and 3A4.
Xenobiotica; the fate of foreign compounds in biological systems (2015-03-27)
Chunhuan Jin, Xin He, Fangliang Zhang, Lina He, Junxiu Chen, Lili Wang, Lijun An, Yaowen Fan
ABSTRACT
1. The present study was conducted to examine the possibility of herb-drug interaction by celastrol, which is a main compound isolated from Tripterygium wilfordii Hook F. using human liver microsomes with cocktail methods. Focused on its inhibitory manner on the metabolism of model probe substrates of five cytochrome P450 isoenzymes (CYP1A2, CYP2C19, CYP2D6, CYP2E1 and CYP3A4) in vitro which are important with the metabolism of different xenobiotics. 2. The results showed that celastrol inhibited the five types of human cytochrome P450 isoforms, with the IC50 values of 2.65 μM (CYP3A4), 5.99 μM (CYP2C19), 6.27 μM (CYP2D6), 7.66 μM (CYP1A2) and 9.38 μM (CYP2E1), respectively. The data indicated that celastrol acted in different manners as an inhibitor of human cytochrome P450 isoforms, which showed that celastrol not only un-competitively inhibited the CYP1A2 and 2E1 activities, but also competitively inhibited the CYP2C19 and 2D6 activities with Ki values of 1.41, 2.29, 5.27 and 4.21 μM, respectively. Celastrol was also a mixed-type inhibitor of CYP3A4, with Ki and Kis values of 2.02 and 5.49 μM, respectively. 3. Celastrol has the potential to inhibit cytochrome P450 activities and may cause the herb-drug interactions. Therefore, the use of celastrol and its preparations with conventional medicines should thus be taken in to account.
Sigma-Aldrich
Phenacetin, ≥98.0% (HPLC)
Sigma-Aldrich
Dextrorphan
Sigma-Aldrich
6-Hydroxychlorzoxazone, ≥98% (HPLC)
Sigma-Aldrich
Potassium phosphate monobasic, BioUltra, Molecular Biology, anhydrous, ≥99.5% (T)
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Acetaminophen, meets USP testing specifications, 98.0-102.0%, powder
Sigma-Aldrich
Chlorzoxazone
Sigma-Aldrich
Potassium phosphate monobasic, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Potassium phosphate monobasic, Molecular Biology, ≥98.0%
Sigma-Aldrich
Potassium phosphate monobasic, ReagentPlus®
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Acetaminophen, BioXtra, ≥99.0%
Sigma-Aldrich
4-Hydroxymephenytoin
Supelco
Acetaminophen solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
4-Hydroxymephenytoin, ≥98% (HPLC)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Acetaminophen, analytical standard
Sigma-Aldrich
Potassium phosphate monobasic, 99.99% trace metals basis
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Celastrol, ≥98% (HPLC), solid