Merck
  • Protective Effects of Quercetin Against HgCl₂-Induced Nephrotoxicity in Sprague-Dawley Rats.
Protective Effects of Quercetin Against HgCl₂-Induced Nephrotoxicity in Sprague-Dawley Rats.
Journal of medicinal food (2015-02-19)
Yu Jin Shin, Jeong Jun Kim, Ye Ji Kim, Won Hee Kim, Eun Young Park, In Young Kim, Han-Seung Shin, Kyeong Seok Kim, Eui-Kyung Lee, Kyu Hyuck Chung, Byung Mu Lee, Hyung Sik Kim
ABSTRACT
Mercury is a well-known environmental pollutant that can cause nephropathic diseases, including acute kidney injury (AKI). Although quercetin (QC), a natural flavonoid, has been reported to have medicinal properties, its potential protective effects against mercury-induced AKI have not been evaluated. In this study, the protective effect of QC against mercury-induced AKI was investigated using biochemical parameters, new protein-based urinary biomarkers, and a histopathological approach. A 250 mg/kg dose of QC was administered orally to Sprague-Dawley male rats for 3 days before administration of mercury chloride (HgCl2). All animals were sacrificed at 24 h after HgCl2 treatment, and biomarkers associated with nephrotoxicity were measured. Our data showed that QC absolutely prevented HgCl2-induced AKI, as indicated by biochemical parameters such as blood urea nitrogen (BUN) and serum creatinine (sCr). In particular, QC markedly decreased the accumulation of Hg in the kidney. Urinary excretion of protein-based biomarkers, including clusterin, kidney injury molecule-1 (KIM-1), neutrophil gelatinase-associated lipocalin (NGAL), monocyte chemoattractant protein-1 (MCP-1), tissue inhibitor of metalloproteinases 1 (TIMP-1), and vascular endothelial growth factor (VEGF) in response to HgCl2 administration were significantly decreased by QC pretreatment relative to that in the HgCl2-treated group. Furthermore, urinary excretion of metallothionein and Hg were significantly elevated by QC pretreatment. Histopathological examination indicated that QC protected against HgCl2-induced proximal tubular damage in the kidney. A TUNEL assay indicated that QC pretreatment significantly reduced apoptotic cell death in the kidney. The administration of QC provided significant protective effects against mercury-induced AKI.
Product Description
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Ammonium chloride, Molecular Biology, suitable for cell culture, ≥99.5%
Sigma-Aldrich
Ammonium chloride, 99.99% trace metals basis
Sigma-Aldrich
Ammonium chloride, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Ammonium chloride, 99.998% trace metals basis
Sigma-Aldrich
Hydrochloric acid solution, for amino acid analysis, ~6 M in H2O
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, Molecular Biology
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Pentostatin, ≥95% (HPLC)
Sigma-Aldrich
Ammonium-14N chloride, 99.99 atom % 14N, 15N-depleted, 99% (CP)
Sigma-Aldrich
5-(N,N-Dimethyl)amiloride hydrochloride
Sigma-Aldrich
Octylamine, 99%
Sigma-Aldrich
Octylamine, ≥99.5%
Sigma-Aldrich
Potassium phosphate monobasic, 99.99% trace metals basis
Sigma-Aldrich
Potassium phosphate monobasic, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Potassium phosphate monobasic, ReagentPlus®
Sigma-Aldrich
Potassium phosphate monobasic, BioUltra, Molecular Biology, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Potassium phosphate monobasic, Molecular Biology, ≥98.0%
Sigma-Aldrich
Uric acid, BioXtra, ≥99% (HPLC)
Sigma-Aldrich
Uric acid, ≥99%, crystalline