Merck
Acid-Sensitive Nanogels for Synergistic Chemo-Photodynamic Therapy.
Macromolecular bioscience (2015-07-15)
Xiaofei Chen, Xuemei Yao, Li Chen, Xuesi Chen
ABSTRACT
Herein, a novel kind of intelligent nanogels have been designed, which can spatiotemporally control the release of drug (doxorubicin) and photosensitizers (porphyrins) to combine chemo-therapy and photodynamic therapy. Further, the fluorescing properties of porphyrins make it possible to be used as a kind of diagnostic bio-imaging agents. In vitro release assays confirm that DOX-loaded nanogels show obvious intracellular pH-sensitive property. The intracellular experiments further prove that drug-loaded nanogels has the significantly antitumor efficacy due to the combination of chemotherapy and photodynamic therapy, which made this drug delivery system hold great potential applications for further cancer treatment.
Sigma-Aldrich
Sodium nitrite, ACS reagent, ≥97.0%
Sigma-Aldrich
Sodium nitrite, 99.999% trace metals basis
Sigma-Aldrich
Sodium nitrite, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium nitrite, anhydrous, Redi-Dri, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.9 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Dimethyl sulfoxide-d6, anhydrous, 99.9 atom % D
Sigma-Aldrich
Dimethyl sulfoxide-d6, "100%", 99.96 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Dimethyl sulfoxide-d6, "Special HOH", ≥99.9 atom % D
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.9 atom % D
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.5 atom % D
Sigma-Aldrich
Dimethyl sulfoxide-d6, "100%", 99.96 atom % D
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.9 atom % D, contains 1 % (v/v) TMS