Merck
  • In vitro affinity reduction of biologic response modifiers from production buffy coat platelets exposed to recombinant protein receptors.
In vitro affinity reduction of biologic response modifiers from production buffy coat platelets exposed to recombinant protein receptors.
Transfusion (2015-03-11)
Per Sandgren, Jenny Rönnmark, Jonas Axelsson
ABSTRACT
Recent studies link biologic response modifiers found in donor platelet (PLT) concentrates to transfusion reactions. We tested a novel method to deplete BRMs from PLT concentrates using apheresis. Whole blood from 25 donors was treated to yield PLTs for in vitro measurements on Days 2, 5, and 7. On Day 7, PLTs were filtrated through columns with either antibody-coated agarose or rh-megalin bound to antibody-coated agarose. In addition, we also tested the naked matrix (agarose) and another apheresis surface containing rh-cubilin bound to agarose. Megalin and cubilin are parts of the protein complex mediating BRM endocytosis in the human kidney. Compared to before filtration (951 × 10(9)  ± 41 × 10(9) cells/L), PLT numbers decreased slightly after filtration over both naked (859 × 10(9)  ± 38 × 10(9) ) and antibody-coated (848 × 10(9)  ± 41 × 10(9) ) matrices (both p < 0.001 vs. before). Concentrations of interleukin (IL)-1β, IL-12 (p40), IL-12 (p70), and IL-7 all decreased by approximately 40% even in the absence of a recombinant surface. After filtration over rh-cubilin, but not rh-megalin, concentrations of IFN-γ, IL-1β, tumor necrosis factor-α, IL-12, and IL-7 all further decreased by 30% to 50%. In a pilot study of in vitro apheresis to deplete BRMs, we found that cell numbers and function remained largely unaffected by filtration. Significant reductions in BRMs occurred already with agarose. However, apheresis with the multiligand receptor rh-cubilin was able to further decrease concentrations.
Sigma-Aldrich
Dipyridamole, ≥98% (HPLC)
Sigma-Aldrich
Calcium chloride, powder, 99.99% trace metals basis
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
PAC-1, ≥98% (HPLC)
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis
Sigma-Aldrich
Calcium chloride
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
Calcium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O