Merck
  • Probiotic Characteristics of Lactobacillus plantarum FH185 Isolated from Human Feces.
Probiotic Characteristics of Lactobacillus plantarum FH185 Isolated from Human Feces.
Korean journal for food science of animal resources (2016-01-14)
Sun-Young Park, Sang-Dong Lim
ABSTRACT
Lactobacillus plantarum FH185 was isolated from the feces of healthy adults. In our previous study, L. plantarum FH185 was demonstrated that it has anti-obesity effect in the in vitro and in vivo test. In order to determine its potential for use as a probiotic, we investigated the physiological characteristics of L. plantarum FH185. The optimum growth temperature of L. plantarum FH185 was 40℃. L. plantarum FH185 showed higher sensitivity to novobiocin in a comparison of fifteen different antibiotics and showed higher resistance to polymyxin B and vancomycin. It also showed higher β-galactosidase and N-acetyl-β-glucosaminidase activities. Moreover, it was comparatively tolerant to bile juice and acid, and inhibited the growths of Salmonella Typhimurium and Staphylococcus aureus with rates of 44.76% and 53.88%, respectively. It also showed high adhesion activity to HT-29 cells compared to L. rhamnosus GG.
Sigma-Aldrich
L-Cysteine, Wacker Chemie AG, ≥98.0%
Sigma-Aldrich
Ampicillin, anhydrous, 96.0-102.0% (anhydrous basis)
Sigma-Aldrich
Chloramphenicol, meets USP testing specifications
Sigma-Aldrich
Chloramphenicol, BioReagent, suitable for plant cell culture
SAFC
L-Cysteine
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Chloramphenicol, ≥98% (HPLC)
Sigma-Aldrich
L-Cysteine, BioUltra, ≥98.5% (RT)
Sigma-Aldrich
L-Cysteine, from non-animal source, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
L-Cysteine, 97%
Sigma-Aldrich
L-Cysteine, ≥97%, FG
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Bacitracin, from Bacillus licheniformis, ≥65 IU/mg
Sigma-Aldrich
Bacitracin, from Bacillus licheniformis, ≥65 IU/mg
Sigma-Aldrich
Ampicillin, meets USP testing specifications
Sigma-Aldrich
Rifampicin, ≥95% (HPLC), powder or crystals
Sigma-Aldrich
Rifampicin, suitable for plant cell culture, BioReagent, ≥95% (HPLC), powder or crystals