Merck
  • Application of pH-sensitive magnetic nanoparticles microgel as a sorbent for the preconcentration of phenoxy acid herbicides in water samples.
Application of pH-sensitive magnetic nanoparticles microgel as a sorbent for the preconcentration of phenoxy acid herbicides in water samples.
Journal of chromatography. A (2015-07-15)
Hadi Tabani, Kamal Khodaei, Yasamin Bide, Farzaneh Dorabadi Zare, Saeed Mirzaei, Ali Reza Fakhari
ABSTRACT
Introducing new sorbents is an interesting and debatable issue in the field of sample preparation. In this study, for the first time, a pH-sensitive magnetic nanoparticles microgel, Fe3O4-SiO2-oly(4-vinylpyridine), was introduced as a new sorbent. The operating mechanism of this sorbent is based on changing the pH value of the sample and consequently the structure of this pH-sensitive microgel is changed. So that, at pH 6.0 the microgel was ready to accept and load the analytes (partial swelling), and when the pH was increased to 8.0, the microgel was closed and analytes were trapped inside the sorbent (deswelling). At pH 2.0 the microgel was opened and the analytes were released from the microgel (swelling). As the adsorption and desorption mechanism is based on changing the pH and only aqueous medium is used as the effluent solvent, this method is introduced as a green extraction method. The use of this microgel resulted in excellent figures of merit. The limits of quantitation and detection for herbicides were obtained within the range of 10-30 and 3-10 ng mL(-1), respectively. Finally, the proposed method was successfully applied to determine the concentration of phenoxy acid herbicides as hazardous materials in water samples.
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Acetic acid, suitable for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, purum, ≥99.0% (TLC)
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%
Sigma-Aldrich
2,4-Dichlorophenoxyacetic acid, 97%
Sigma-Aldrich
2,4-Dichlorophenoxyacetic acid, ≥95%, crystalline