Merck
  • Mercapto functionalized silica entrapped polyacrylamide hydrogel: Arsenic adsorption behaviour from aqueous solution.
Mercapto functionalized silica entrapped polyacrylamide hydrogel: Arsenic adsorption behaviour from aqueous solution.
Journal of colloid and interface science (2015-07-08)
Rajesh Kumar, S K Jain, S Verma, P Malodia
ABSTRACT
In this article, 3-mercaptopropyl functionalized silica entrapped polyacrylamide hydrogel (MPFS-PAA) was prepared and characterized by FT-IR, scanning electron microscopy (SEM) and energy dispersion X-ray spectroscopy (EDS). Synthesized hydrogel was evaluated for removal of arsenic(III) from aqueous solution. Adsorption studies were carried out by batch method as function of contact time, initial concentration of arsenic and pH. As(III) adsorption data fitted well with Langmuir and Freundlich isotherm models. Adsorption capacity of arsenic 92.5 μg/g was obtained at initial concentration of 100 μg/L by Langmuir isotherm. Adsorption kinetics was tested for pseudo-second order reaction at different contact time. The rate constants of pseudo second order reaction were calculated and good correlation coefficient R(2) 99.67 obtained. The results indicates that MPFS-PAA is an effective adsorbent for removal of As(III) from aqueous solution.
Sigma-Aldrich
VenPure® SF, powder
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
Sodium borohydride solution, 2.0 M in triethylene glycol dimethyl ether
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
Sodium borohydride solution, ~12 wt. % in 14 M NaOH
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
Hydrochloric acid solution, for amino acid analysis, ~6 M in H2O
Sigma-Aldrich
Potassium iodide, AnhydroBeads, −10 mesh, 99.998% trace metals basis
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Potassium iodide, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium iodide, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Potassium iodide, ≥99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, Molecular Biology
Sigma-Aldrich
Potassium iodide, suitable for plant cell culture
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis