Merck
Sphingomonas flavus sp. nov. isolated from road soil.
Archives of microbiology (2015-05-27)
Juan Du, Hina Singh, KyungHwa Won, Jung-Eun Yang, Shahina Akter, Feng-Xie Jin, Tae-Hoo Yi
ABSTRACT
A yellow-colored, Gram-negative, strictly aerobic, non-motile, rod-shaped bacterium, designated THG-MM5(T), was isolated from road soil in Yongin-si, Gyeonggi-do, Republic of Korea. Based on 16S rRNA gene sequence, strain THG-MM5(T) was moderately related to Sphingomonas sediminicola KACC 15039(T) (96.1%), Sphingomonas ginsengisoli KACC 16858(T) (96.1%) and Sphingomonas jaspsi KACC 13230(T) (96.0%). Chemotaxonomic data revealed that strain THG-MM5(T) possesses ubiquinone-10 as the only respiratory quinone, sym-homospermidine as the major polyamine and summed feature 3 (C16:1 ω7c and/or C16:1 ω6c), C18:1 ω7c and C16:0 as the major fatty acids. The polar lipid profile included sphingoglycolipid. The DNA G + C content was 60.7 mol%. These data, together with phenotypic characterization, corroborated the affiliation of strain THG-MM5(T) to the genus Sphingomonas. Thus, the isolate represents a novel species, for which the name Sphingomonas flavus sp. nov. is proposed, with THG-MM5(T) as the type strain (=KACC 18277(T) = CCTCC AB 2014320(T)).
Sigma-Aldrich
Calcium chloride, powder, 99.99% trace metals basis
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
Calcium chloride
Sigma-Aldrich
Trichostatin A, ≥98% (HPLC), from Streptomyces sp.
Sigma-Aldrich
Deoxyribonucleic acid sodium salt from Escherichia coli strain B, Genomic, unsheared
Sigma-Aldrich
Calcium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Phosphomolybdic acid solution, spray reagent, 10% in ethanol
Sigma-Aldrich
L-Tyrosine, FG
Sigma-Aldrich
Phosphomolybdic acid solution, 20 wt. % in ethanol
Sigma-Aldrich
L-Tyrosine, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
Sulfuric acid, 99.999%
Supelco
Phosphomolybdic acid hydrate, ≥99.99% trace metals basis
SAFC
L-Tyrosine
Sigma-Aldrich
L-Tyrosine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
Ferric citrate, BioReagent, suitable for cell culture
Sigma-Aldrich
Iron(III) citrate, technical grade
Sigma-Aldrich
Phosphomolybdic Acid Solution
Sigma-Aldrich
L-Tyrosine, reagent grade, ≥98% (HPLC)
SAFC
Ferric citrate