Merck
A new delivery system for auristatin in STxB-drug conjugate therapy.
European journal of medicinal chemistry (2015-04-08)
Cornélie Batisse, Estelle Dransart, Rafik Ait Sarkouh, Laura Brulle, Siau-Kun Bai, Sylvie Godefroy, Ludger Johannes, Frédéric Schmidt
ABSTRACT
A key challenge in anticancer therapy is to gain control over the biodistribution of cytotoxic drugs. The most promising strategy consists in conjugating drugs to tumor-targeting carriers, thereby combining high cytotoxic activity and specific delivery. To target Gb3-positive cancer cells, we exploit the non-toxic B-subunit of Shiga toxin (STxB). Here, we have conjugated STxB to highly potent auristatin derivatives (MMA). A former linker was optimized to ensure proper drug-release upon reaching reducing environments in target cells, followed by a self-immolation step. Two conjugates were successfully obtained, and in vitro assays demonstrated the potential of this targeting system for the selective elimination of Gb3-positive tumors.
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis