Merck
  • Development of carboxymethyl cellulose nonwoven sheet as a novel hemostatic agent.
Development of carboxymethyl cellulose nonwoven sheet as a novel hemostatic agent.
Journal of bioscience and bioengineering (2014-12-10)
Seiichi Ohta, Toru Nishiyama, Megumu Sakoda, Kyoko Machioka, Masaya Fuke, Shigetoshi Ichimura, Fuyuki Inagaki, Atsushi Shimizu, Kiyoshi Hasegawa, Norihiro Kokudo, Makoto Kaneko, Yutaka Yatomi, Taichi Ito
ABSTRACT
Carboxymethyl cellulose (CMC) is a plant-derived material that has high biocompatibility and water solubility. We developed a CMC nonwoven sheet as a hemostatic agent by carboxymethylating a continuous filament cellulose nonwoven sheet. The CMC nonwoven sheet was able to absorb water and dissolve in it. The rates of absorption and dissolution depended on the degree of carboxymethylation. After dissolving in blood, CMC accelerated clot development (possibly owing to the incorporation of CMC into fibrin fibers) and increased the viscosity of the blood, both of which would contribute to the improved blood clotting of an injured surface. In vivo experiments using a rat tail cutting method showed that a CMC nonwoven sheet shortened the bleeding time of the tail when applied to the cut surface. The hemostatic effect of the CMC nonwoven sheet was almost at the same level as a commercial hemostatic bandage. These results suggest that a CMC nonwoven sheet could be used as a novel sheet-type hemostatic agent.
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Acetic acid, suitable for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Citrate Concentrated Solution, BioReagent, suitable for coagulation assays, 4 % (w/v)
Sigma-Aldrich
Citrate Concentrated Solution, BioUltra, Molecular Biology, 1 M in H2O
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Calcium chloride, powder, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Calcium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, purum, ≥99.0% (TLC)
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
Calcium chloride
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis