Merck
  • Synthesis and Cytotoxic Evaluation of Pyran, Dihydropyridine and Thiophene Derivatives of 3-Acetylcoumarin.
Synthesis and Cytotoxic Evaluation of Pyran, Dihydropyridine and Thiophene Derivatives of 3-Acetylcoumarin.
Chemical & pharmaceutical bulletin (2015-09-04)
Rafat Milad Mohareb, Nadia Youssef Megally Abdo
ABSTRACT
A series of coumarin analogues bearing 4H-pyran rings 2a-d, 11a-d and 1,4-dihydropyridine rings 3a-d, 12a-d at position 3 were synthesized starting from either 3-acetyl coumarin (1) or the coumarin acetohydrazide derivative 4. Condensation of 3-acetylcoumarin (1) with 2-cyanoacetohydrazide afforded 2-cyano-N'-{1-[2-oxo-2H-chromen-3-yl]ethylidene}acetohydrazide (4). Reaction of compound 4 with elemental sulfur and either malononitrile or ethyl cyanoacetate afforded the thiophene derivatives 8 and 9, respectively. The structures of the newly synthesized compounds were confirmed on the basis of their spectral data and elemental analyses. All synthesized compounds were screened for their in vitro anticancer activity against six human cancer cell lines and normal fibroblasts. Several compounds showed potent inhibition with an IC50 value of ˂870 nM. Compound 3d exhibited equivalent cytotoxic effect as the standard CHS 828 against a breast cancer cell line (IC50 value=18 nM). Normal fibroblast cells (WI38) were affected to a much lesser extent (IC50 value >10000 nM).
Sigma-Aldrich
Sulforhodamine B sodium salt, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Sulforhodamine B sodium salt, Technical grade
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
L-Glutamine
SAFC
L-Glutamine
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Cholesteryl hemisuccinate tris salt, anionic detergent
Sigma-Aldrich
Sulforhodamine B, Dye content 75 %