Merck
  • pH value manipulated phase transition, microstructure evolution and tunable upconversion luminescence in Yb(3+)-Er(3+) codoped LiYF4/YF3 nanoparticles.
pH value manipulated phase transition, microstructure evolution and tunable upconversion luminescence in Yb(3+)-Er(3+) codoped LiYF4/YF3 nanoparticles.
Dalton transactions (Cambridge, England : 2003) (2015-08-06)
Song Ye, Rongxuan Hu, Nan Jiang, Huiyun Wang, Deping Wang
ABSTRACT
The pH value plays an important role in controlling the crystallization process and microstructure of the final products in the synthesis of nanocrystals with a solvothermal method. This work reported the effect of the mother solution pH value on the precipitation of LiYF4 and YF3 nanoparticles, as well as the microstructure evaluation of YF3 from a bowknot-like to spindle-like shape. Spectroscopy study suggests that there is strong correlation between the upconversion emission properties of the Yb(3+)-Er(3+) couple and the phase and the microstructure of the host. The strongest emissions and lowest red-to-green ratio are observed in the bowknot-like YF3 nanocrystals with the largest open ends. Further spectral investigation indicates that the phase and microstructure dependent upconversion properties are associated with the upconversion efficiency. The present study is of great importance in the design and synthesis of rare earth ion doped nanocrystals with tunable upconversion properties.
Sigma-Aldrich
Oleic acid, natural, FCC
Sigma-Aldrich
Oleic acid, meets analytical specification of Ph, Eur., 65.0-88.0% (GC)
Sigma-Aldrich
Nitric acid, 70%, purified by redistillation, ≥99.999% trace metals basis
Sigma-Aldrich
Nitric acid, ACS reagent, ≥90.0%
Sigma-Aldrich
Oleic acid, technical grade, 90%
Sigma-Aldrich
Oleic acid, BioReagent, suitable for cell culture
Sigma-Aldrich
Oleic acid, ≥99% (GC)
Sigma-Aldrich
Nitric-14N acid solution, ~10 N in H2O, 99.99 atom % 14N
Sigma-Aldrich
Ammonium fluoride, ≥99.99% trace metals basis