Merck
  • Derivatives containing both coumarin and benzimidazole potently induce caspase-dependent apoptosis of cancer cells through inhibition of PI3K-AKT-mTOR signaling.
Derivatives containing both coumarin and benzimidazole potently induce caspase-dependent apoptosis of cancer cells through inhibition of PI3K-AKT-mTOR signaling.
Anti-cancer drugs (2015-03-27)
Haitao Liu, Yubin Wang, Ashok Sharma, Rui Mao, Na Jiang, Boying Dun, Jin-Xiong She
ABSTRACT
Coumarins are a large family of compounds derived from a wide range of plants, fungi, and bacteria, and coumarin derivatives can have extremely variable structures and consequently diverse biological properties including antitumor activity. Compounds that bear a benzimidazole moiety are known to possess antitumor activity and a variety of other biological activities. High-throughput screening of a compound library identified a coumarin-containing and a benzimidazole-containing compound [#32, 7-(diethylamino)-3-(1-methyl-1H-benzimidazol-2-yl)-2H-chromen-2-one] that has potent anticancer activity. Evaluation of 17 additional analogs further identified three compounds with anticancer activity in 14 different human cancer cell lines. Fluorescence-activated cell sorting and western blotting analyses suggested that these compounds can induce caspase-dependent apoptosis. Real-time reverse transcriptase PCR analyses of 26 cancer-related genes revealed that seven genes (NPPB, ATF3, DDIT4, CDH10, TSPAN14, TXNIP, and AXL) were significantly upregulated and nine genes (PAGE4, LRP8, SNCAIP, IGFBP5, SLCO2A1, CLDN2, ESRRG, D2HGDH, and PDGFRA) were significantly downregulated. The most upregulated gene is natriuretic peptide precursor B (NPPB) or brain natriuretic peptide, which is increased by 7-, 27-, and 197-fold at 12, 24, and 48 h, respectively. The second most upregulated gene is ATF3, which is increased by 23-fold at the 48 h timepoint. PAGE4 and IGFBP5 are the two most downregulated genes, with a 17-fold reduction in both genes. The expression of several genes (DDIT4, PDGFRA, LRP8, IGFBP5) and western blotting data on key signaling proteins indicate that compound #32 significantly inhibits the PI3K-AKT-mTOR pathway, an intracellular signaling pathway critical in cell proliferation and apoptosis.
Sigma-Aldrich
Coumarin, ≥99% (HPLC)
Sigma-Aldrich
Tris(tert-butoxy)silanol, 99.999%
Sigma-Aldrich
Benzimidazole, 98%