Merck
  • Emulsification and antioxidation of biosurfactant extracts from Chinese medicinal herbs fermentation in vitro.
Emulsification and antioxidation of biosurfactant extracts from Chinese medicinal herbs fermentation in vitro.
Journal of bioscience and bioengineering (2015-03-31)
Chunyeh Chen, Tachen Lin, Youmin Shieh
ABSTRACT
Much attention has been paid to biosurfactants produced using microorganisms, but little direct evidence for the development of natural biosurfactants combined with Chinese medicinal herbs are available. We investigated the emulsification and antioxidation of biosurfactant extracts from Chinese medicinal herb fermentation (BECMHF) in vitro and their application in water retention capacity and the skin prick and allergy test (SPAT) index for skin cells. The results showed that the water retention capacity of BECMHF was positively associated with the emulsification index. The SPAT index of 8 Chinese medicinal herbs was 0 at a 1% or 2% concentration, suggesting no sensitivity or adverse effects on the skin cells. Eight BECMHFs produced using Alcaligenes piechaudii CC-ESB2 exhibited antioxidant capabilities, including 2,2-diphenyl-1-picrylhydrazyl (DPPH) radical and superoxide scavenging activity, and superoxide dismutase (SOD)-like activity at a concentration of 10 mg/ml. The mechanism involved inhibitory effects on nitrite, inducible nitric oxide synthase (iNOS) expression, and reactive oxygen species (ROSs) generation. BECMHFs exhibit favorable antioxidative properties in health food and satisfactory emulsifying and moisturizing characteristics in cosmetic formulations, which have potential applications in the health food and cosmetic industries, respectively.
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
Nitrotetrazolium Blue chloride, powder, electrophoresis grade
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
Nitrotetrazolium Blue chloride, ≥90.0% (HPLC)
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Chloroform, biotech. grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Chloroform, contains amylenes as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, meets analytical specification of BP, 99-99.4% (GC)
Supelco
L-Ascorbic acid, analytical standard
Sigma-Aldrich
Chloroform, ReagentPlus®, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%