Merck
  • Partial protection and abomasal cytokine expression in sheep experimentally infected with Haemonchus contortus and pre-treated with Taenia hydatigena vesicular concentrate.
Partial protection and abomasal cytokine expression in sheep experimentally infected with Haemonchus contortus and pre-treated with Taenia hydatigena vesicular concentrate.
Veterinary parasitology (2015-05-12)
J A Buendía-Jiménez, M A Muñoz-Guzmán, M A Vega-López, C Cuenca-Verde, J P Martínez-Labat, J A Cuéllar-Ordaz, F Alba-Hurtado
ABSTRACT
The abomasal expression of IL-2, IL-4, IL-6, IL-10 and IFNγ in lambs experimentally infected with Haemonchus contortus and its relationship to protection induced by a Taenia hydatigena larvae vesicular concentrate (ThLVC) were evaluated. The lambs that were only infected with H. contortus larvae showed a worm burden greater (p<0.05) than the lambs that received ThLVC prior to infection. Moreover, the lambs that received ThLVC showed a greater (p<0.05) number of blood eosinophils than the lambs that did not receive the ThLVC. In general, the lambs that received ThLVC prior to infection had a greater amount of eosinophils and mast cells and higher in situ expression of IFNγ, IL-2, IL-4, IL-6 and IL-10 in the abomasal wall than the lambs that were infected with H. contortus only or that received ThLVC (p<0.05) only. A higher expression of IL-2 and IFNγ in the submucosa compared to the abomasal mucosa and a higher expression of IL-4 in the abomasal mucosa compared to the submucosa was observed (p<0.05). These results suggest that there is a Th1 type response in the abomasal submucosa and a Th2 type response in in the abomasal mucosa. The amount of eosinophils and mast cells and the in situ expression of IFNγ, IL-2, IL-4 and IL-6 in the abomasal walls were negatively correlated with the worm burden (p<0.05). These results suggest that ThLVC is a non-specific immune stimulator for the abomasal immune response, and it is likely that the protection observed is the result of this effect.
Sigma-Aldrich
Iodoacetamide, ≥99% (NMR), crystalline
Sigma-Aldrich
Chromotrope 2R, suitable for modified Gomori Trichrome stain
Sigma-Aldrich
Iodoacetamide, BioUltra
Sigma-Aldrich
Iodoacetamide, Single use vial of 56 mg
SAFC
Iodoacetamide
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)