Merck
  • In vitro and in vivo antioxidant activity of polyphenols extracted from black highland barley.
In vitro and in vivo antioxidant activity of polyphenols extracted from black highland barley.
Food chemistry (2015-10-17)
Yingbin Shen, Hui Zhang, Liling Cheng, Li Wang, Haifeng Qian, Xiguang Qi
ABSTRACT
The objective of this study was to determine the antioxidant capacity of polyphenols extracted from black highland barley (BHLPE) in vitro and in vivo. BHLPE was found to have strong superoxide radical, hydroxyl radical and 2,2-diphenyl-1-picrylhydrazyl radical-scavenging activity, ferric reducing antioxidant power and moderate metal ion-chelating activity. Compared with a high fat diet (HFD) group, mice that were administered 600mg BHLPE/kg body weight showed significant decreases in total cholesterol, low-density lipoprotein cholesterol and the atherosclerosis index, in addition to markedly increased high-density lipoprotein cholesterol levels. Furthermore, the antioxidant defense system and antioxidant gene expression were significantly improved in vivo in mice that were administered BHLPE compared with mice in the HFD group. These results suggest that BHLPE has significant potential as a natural antioxidant to promote health and to reduce the risk of disease.
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%
Sigma-Aldrich
Phenazine methosulfate
Sigma-Aldrich
trans-Ferulic acid, ≥99%
Sigma-Aldrich
trans-Ferulic acid, 99%
Sigma-Aldrich
Gallic acid, 97.5-102.5% (titration)
Sigma-Aldrich
(±)-6-Hydroxy-2,5,7,8-tetramethylchromane-2-carboxylic acid, 97%
Supelco
2,4,6-Tris(2-pyridyl)-s-triazine, ≥98%
Sigma-Aldrich
Nitrotetrazolium Blue chloride, ≥90.0% (HPLC)
Sigma-Aldrich
Nitrotetrazolium Blue chloride, powder, electrophoresis grade
Sigma-Aldrich
p-Coumaric acid, ≥98.0% (HPLC)