Merck
  • High Performance Non-enzymatic Glucose Sensor Based on One-Step Electrodeposited Nickel Sulfide.
High Performance Non-enzymatic Glucose Sensor Based on One-Step Electrodeposited Nickel Sulfide.
Chemistry (Weinheim an der Bergstrasse, Germany) (2015-05-23)
Padmanathan Karthick Kannan, Chandra Sekhar Rout
ABSTRACT
Nanostructured NiS thin film was prepared by a one-step electrodeposition method and the structural, morphological characteristics of the as-prepared films were analyzed by X-ray diffractometry (XRD), field emission scanning electron microscopy (FESEM) and energy dispersive X-ray analysis (EDAX). The electrocatalytic activity of NiS thin film towards glucose oxidation was investigated by fabricating a non-enzymatic glucose sensor and the sensor performance was studied by cyclic voltammetry (CV) and amperometry. The fabricated sensor showed excellent sensitivity and low detection limit with values of 7.43 μA μM(-1)  cm(-2) and 0.32 μM, respectively, and a response time of <8 s.
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
Thioacetamide, reagent grade, 98%
Sigma-Aldrich
Thioacetamide, ACS reagent, ≥99.0%