Merck
  • Chemical Compositions and Biological Activities of Essential Oils of Beilschmiedia glabra.
Chemical Compositions and Biological Activities of Essential Oils of Beilschmiedia glabra.
Natural product communications (2015-09-29)
Wan Mohd Nuzul Hakimi Wan Salleh, Farediah Ahmada, Khong Heng Yen, Razauden Mohamed Zulkifli
ABSTRACT
This study was designed to examine the chemical compositions of essential oils from Beilschmiedia glabra and their antioxidant, antimicrobial, antityrosinase, acetylcholinesterase and anti-inflammatory activities. In total, 47 components were identified in the essential oils, which made up 86.8% and 89.7% of the leaf and bark oils, respectively. The leaf oil is composed mainly of β-eudesmol (15.4%), β-selinene (12.2%), caryophyllene oxide (8.1%) and γ-gurjunene (5.2%), while the bark oil contains high percentages of β-eudesmol (19.3%), β-selinene (16.9%), δ-cadinene (15.8%), germacrene D (9.8%) and β-caryophyllene (5.5%). Antioxidant activity showed that the leaf oil has the highest phenolic content at 233.4 mg GA/g, while the bark oil showed potent activity in the β- carotene/linoleic acid bleaching assay. However, both oils showed weak activity in the DPPH and ABTS assays. For antimicrobial activity, the leaf and bark oils displayed strong activity against Candida glabrata and Saccharomyces cerevisiae with MIC values of 31.3 and 62.5 μg/mL, respectively. Percentage inhibitions against tyrosinase (leaf 73.7%; bark 76.0%) and acetylcholinesterase (leaf 48.1%; bark 45.2%) were tested at a concentration of 1 mg/mL, while anti-inflammatory activity (leaf 59.7%; bark 48.9%) was evaluated at a concentration of 100 μM. Evaluation of these assays indicated moderate levels of activity.
Sigma-Aldrich
Acetylthiocholine iodide, ≥98% (TLC), powder or crystals
Sigma-Aldrich
2,2-Diphenyl-1-picrylhydrazyl
Sigma-Aldrich
α-Linoleic acid, ≥98%
Sigma-Aldrich
Acetylthiocholine iodide, ≥99.0% (AT)
Sigma-Aldrich
Sodium carbonate-12C, 99.9 atom % 12C
Sigma-Aldrich
2,6-Di-tert-butyl-4-methylphenol, ≥99.0% (GC), powder
Sigma-Aldrich
3,4-Dihydroxy-L-phenylalanine, ≥98% (TLC)
Sigma-Aldrich
Potassium persulfate, 99.99% trace metals basis
Sigma-Aldrich
Kojic acid
Sigma-Aldrich
Dextrose, 97.5-102.0% anhydrous basis, meets EP, BP, JP, USP testing specifications
Sigma-Aldrich
Gallic acid, 97.5-102.5% (titration)
Sigma-Aldrich
Sodium sulfate, BioXtra, ≥99.0%
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
5,5′-Dithiobis(2-nitrobenzoic acid), ≥98%, BioReagent, suitable for determination of sulfhydryl groups
Sigma-Aldrich
Sodium sulfate, BioUltra, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Sodium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Sodium carbonate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium carbonate, BioUltra, anhydrous, ≥99.5% (calc. on dry substance, T)
Sigma-Aldrich
Sodium carbonate, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
5,5′-Dithiobis(2-nitrobenzoic acid), ReagentPlus®, 99%
Sigma-Aldrich
Linoleic acid, technical, 58-74% (GC)
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbic acid, reagent grade