Merck
  • Peptide Bioconjugates of Electron-Poor Metallocenes: Synthesis, Characterization, and Anti-Proliferative Activity.
Peptide Bioconjugates of Electron-Poor Metallocenes: Synthesis, Characterization, and Anti-Proliferative Activity.
Chembiochem : a European journal of chemical biology (2015-05-28)
Marcus Maschke, Jens Grohmann, Claudia Nierhaus, Max Lieb, Nils Metzler-Nolte
ABSTRACT
We report the synthesis of metallocene compounds Cp2 M with two different electron-withdrawing substituents on both cyclopentadienyl rings (hexafluoroacetone (HFA) and chlorobenzoyl (1-5); HFA and COOH (6 and 7), M=Fe or Ru). The COOH-containing derivatives were used to synthesize peptide bioconjugates with enkephalin (8 and 9) and neurotensin (10 and 11) as well as fluorescein-labeled neurotensin (12). All the molecules were fully characterized, including X-ray structures for 6 and 7. The physicochemical properties (lipophilicity and electrochemistry) and cytotoxicity on MCF-7, HT-29, and PT-45 cancer cells were evaluated for selected compounds. Electrochemical investigation by cyclic voltammetry revealed that all bis-substituted metallocenes are up to 300 mV harder to oxidize compared to the monosubstituted 2-ferrocenylhexafluoropropan-2-ol (FcHFA: Δ${E{{{\rm f}\hfill \atop 0\hfill}}}$=214 mV; disubstituted derivatives: up to Δ${E{{{\rm f}\hfill \atop 0\hfill}}}$=512 mV; both vs. FcH(0/+) ). For the bis-substituted compounds, log P determinations by RP-HPLC showed increased lipophilicity in comparison to the monosubstituted FcHFA and RcHFA. Cellular uptake was investigated by fluorescence microcopy, and this revealed endosomal entrapment for 12.
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
BES, ≥99.0% (titration)
Sigma-Aldrich
Potassium chloride, Molecular Biology, ≥99.0%
Sigma-Aldrich
Potassium chloride, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Potassium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Potassium chloride, 99.999% trace metals basis
Sigma-Aldrich
Potassium chloride, ≥99.99% trace metals basis
Sigma-Aldrich
Potassium chloride, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Potassium chloride solution, 0.075 M, sterile-filtered, BioXtra, suitable for cell culture