Merck
  • Detection of tetrahydrocannabinol residues on hands by ion-mobility spectrometry (IMS). Correlation of IMS data with saliva analysis.
Detection of tetrahydrocannabinol residues on hands by ion-mobility spectrometry (IMS). Correlation of IMS data with saliva analysis.
Analytical and bioanalytical chemistry (2015-06-15)
Saskia Sonnberg, Sergio Armenta, Salvador Garrigues, Miguel de la Guardia
ABSTRACT
Ion-mobility spectroscopy (IMS) was evaluated as a high-throughput, cheap, and efficient analytical tool for detecting residues of tetrahydrocannabinol (THC) on hands. Regarding the usefulness of hand residues as potential samples for determining THC handling and abuse, we studied the correlation between data obtained from cannabis consumers who were classified as positive after saliva analysis and from those who were classified as positive on the basis of the information from hand-residue analysis. Sampling consisted of wiping the hands with borosilicate glass microfiber filters and introducing these directly into the IMS after thermal desorption. The possibility of false positive responses, resulting from the presence of other compounds with a similar drift time to THC, was evaluated and minimised by applying the truncated negative second-derivative algorithm. The possibility of false negative responses, mainly caused by competitive ionisation resulting from nicotine, was also studied. Graphical abstract THC residues: from hands to analytical signals.
Sigma-Aldrich
Poly(tetrafluoroethylene), powder (free-flowing), 1 μm particle size
Sigma-Aldrich
Poly(tetrafluoroethylene), powder (free-flowing), ≤12 μm particle size
Sigma-Aldrich
Poly(tetrafluoroethylene), powder, 35 μm particle size
Sigma-Aldrich
Poly(tetrafluoroethylene), powder, >40 μm particle size
Sigma-Aldrich
Poly(tetrafluoroethylene), beads
Sigma-Aldrich
Poly(tetrafluoroethylene), powder, 200 μm particle size
Sigma-Aldrich
Poly(tetrafluoroethylene), powder, ≥350 μm particle size