Merck
  • Vitamin B12 modulates the transcriptome of the skin microbiota in acne pathogenesis.
Vitamin B12 modulates the transcriptome of the skin microbiota in acne pathogenesis.
Science translational medicine (2015-06-26)
Dezhi Kang, Baochen Shi, Marie C Erfe, Noah Craft, Huiying Li
ABSTRACT
Various diseases have been linked to the human microbiota, but the underlying molecular mechanisms of the microbiota in disease pathogenesis are often poorly understood. Using acne as a disease model, we aimed to understand the molecular response of the skin microbiota to host metabolite signaling in disease pathogenesis. Metatranscriptomic analysis revealed that the transcriptional profiles of the skin microbiota separated acne patients from healthy individuals. The vitamin B12 biosynthesis pathway in the skin bacterium Propionibacterium acnes was significantly down-regulated in acne patients. We hypothesized that host vitamin B12 modulates the activities of the skin microbiota and contributes to acne pathogenesis. To test this hypothesis, we analyzed the skin microbiota in healthy subjects supplemented with vitamin B12. We found that the supplementation repressed the expression of vitamin B12 biosynthesis genes in P. acnes and altered the transcriptome of the skin microbiota. One of the 10 subjects studied developed acne 1 week after vitamin B12 supplementation. To further understand the molecular mechanism, we revealed that vitamin B12 supplementation in P. acnes cultures promoted the production of porphyrins, which have been shown to induce inflammation in acne. Our findings suggest a new bacterial pathogenesis pathway in acne and provide one molecular explanation for the long-standing clinical observation that vitamin B12 supplementation leads to acne development in a subset of individuals. Our study discovered that vitamin B12, an essential nutrient in humans, modulates the transcriptional activities of skin bacteria, and provided evidence that metabolite-mediated interactions between the host and the skin microbiota play essential roles in disease development.
Sigma-Aldrich
Cyanocobalamin, meets USP testing specifications
Sigma-Aldrich
Vitamin B12, ≥98%
Sigma-Aldrich
Vitamin B12, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥98%