Merck
  • Competitive Solubilization of Cholesterol/Cholesteryl Oleate and Seven Species of Sterol/Stanol in Model Intestinal Solution System.
Competitive Solubilization of Cholesterol/Cholesteryl Oleate and Seven Species of Sterol/Stanol in Model Intestinal Solution System.
Journal of oleo science (2015-07-03)
Keisuke Matsuoka, Asumi Kase, Takashi Matsuo, Yusuke Ashida
ABSTRACT
The addition of plant sterols/stanols (sterols or stanols) can reduce the solubilization of cholesterol in a model intestinal solution system. We studied the molecular structure of seven different sterols/stanols and the effect they had on the solubilization of cholesterol or cholesterol ester in a model intestinal solution. The differences in the molecular structures of the sterol/stanol species influenced their abilities to reduce the solubility of cholesterol in the competitive solubilization experiments. Cholestanol whose molecular structure resembled cholesterol was the most effective at reducing the solubilization of cholesterol and cholesterol ester, with the solubilities of cholesterol and cholesteryl oleate being 41% and 39% respectively of the values observed for the single solubilizate systems. β-Sitosterol was also able to reduce the solubilities of cholesterol and cholesteryl oleate to 43% and 45% of those observed in a single solubilizate system. Both, stigmasterol and brassicasterol have an unsaturated double bond in a steroid side chain and did not exhibit major cholesterol-lowering effects. These results were reflected by the Gibbs free energy change values (ΔG(0)) for solubilization, where the sterol/stanol species with cholesterol-lowering effects had similar or larger negative ΔG(0) values than those observed for cholesterol.
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Naphthalene, suitable for scintillation, ≥99%
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Sigma-Aldrich
Naphthalene, 99%
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
Pyrene, sublimed grade, 99%
Sigma-Aldrich
Pyrene, puriss. p.a., suitable for fluorescence, ≥99.0% (GC)
Sigma-Aldrich
Pyrene, 98%
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications
Sigma-Aldrich
Cholesteryl oleate, ≥98% (HPLC; detection at 205 nm)
Sigma-Aldrich
Sodium taurochenodeoxycholate
Sigma-Aldrich
Oleic acid, BioReagent, suitable for cell culture
Sigma-Aldrich
Oleic acid, ≥99% (GC)
Sigma-Aldrich
Oleic acid, technical grade, 90%
Sigma-Aldrich
Oleic acid, meets analytical specification of Ph, Eur., 65.0-88.0% (GC)
Sigma-Aldrich
Oleic acid, natural, FCC