Merck
  • In vitro evaluation of different methods of handling human liposuction aspirate and their effect on adipocytes and adipose derived stem cells.
In vitro evaluation of different methods of handling human liposuction aspirate and their effect on adipocytes and adipose derived stem cells.
Journal of cellular physiology (2015-03-05)
Paola Palumbo, Gianfranca Miconi, Benedetta Cinque, Cristina La Torre, Francesca Lombardi, Giovanni Zoccali, Gino Orsini, Pietro Leocata, Maurizio Giuliani, Maria Grazia Cifone
ABSTRACT
Nowadays, fat tissue transplantation is widely used in regenerative and reconstructive surgery. However, a shared method of lipoaspirate handling for ensuring a good quality fat transplant has not yet been established. The study was to identify a method to recover from the lipoaspirate samples the highest number of human viable adipose tissue-derived stem cells (hADSCs) included in stromal vascular fraction (SVF) cells and of adipocytes suitable for transplantation, avoiding an extreme handling. We compared the lipoaspirate spontaneous stratification (10-20-30 min) with the centrifugation technique at different speeds (90-400-1500 × g). After each procedure, lipoaspirate was separated into top oily lipid layer, liquid fraction, "middle layer", and bottom layer. We assessed the number of both adipocytes in the middle layer and SVF cells in all layers. The histology of middle layer and the surface phenotype of SVF cells by stemness markers (CD105+, CD90+, CD45-) was analyzed as well. The results showed a normal architecture in all conditions except for samples centrifuged at 1500 × g. In both methods, the flow cytometry analysis showed that greater number of ADSCs was in middle layer; in the fluid portion and in bottom layer was not revealed significant expression levels of stemness markers. Our findings indicate that spontaneous stratification at 20 min and centrifugation at 400 × g are efficient approaches to obtain highly viable ADSCs cells and adipocytes, ensuring a good thickness of lipoaspirate for autologous fat transfer. Since an important aspect of surgery practice consists of gain time, the 400 × g centrifugation could be the recommended method when the necessary instrumentation is available.
Sigma-Aldrich
Adenosine
Sigma-Aldrich
Adenosine, ≥99%
Sigma-Aldrich
Adenosine, BioReagent, suitable for cell culture
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Trypan Blue solution, 0.4%, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Rabbit Anti-Goat IgG Antibody, HRP conjugate, 1 mg/mL (after reconstitution), Chemicon®
Sigma-Aldrich
Trypan Blue, ≥80% (HPLC), Dye content 60 %
Sigma-Aldrich
Aphidicolin from Nigrospora sphaerica, ≥98% (HPLC), powder
Sigma-Aldrich
Trypan Blue, powder, BioReagent, suitable for cell culture