Merck
  • Intracellular plasma membrane guidance of Photorhabdus asymbiotica toxin is crucial for cell toxicity.
Intracellular plasma membrane guidance of Photorhabdus asymbiotica toxin is crucial for cell toxicity.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2015-03-19)
Thomas Jank, Christoph Trillhaase, Natalia Brozda, Marcus Steinemann, Carsten Schwan, Regine Süss, Klaus Aktories
ABSTRACT
The bacterial toxin Photorhabdus asymbiotica toxin (PaTox) modifies Rho proteins by tyrosine GlcNAcylation and heterotrimeric Gα proteins by deamidation. Inactivation of Rho proteins results in F-actin disassembly in host cells. Here, we analyzed the subcellular distribution of PaTox and show that the glycosyltransferase domain of PaTox associates with the negatively charged inner surface of the plasma membrane. Localization studies with site-directed mutants, liposome precipitation analysis, lipid overlay assays, and confocal time-lapse microscopy revealed that a patch of positively charged lysine and arginine residues located on helix α1 of the glycosyltransferase is essential for membrane attachment. Using a helix1 deletion mutant, we show that plasma membrane localization of PaTox is essential for cytotoxicity and proved this by substitution of helix1 by an N-terminal myristoylation signal peptide, which restored plasma membrane localization and cytotoxicity. Furthermore, we also show that the intracellular deamidase activity of PaTox depends on the presence of the membrane localization domain. Comparison of PaTox membrane-binding domain with the 4-helix-bundle membrane-binding domain of Pasteurella multocida toxin, Vibrio cholerae multifunctional autoprocessing repeats-in-toxin, and clostridial glucosylating toxins revealed similar spatial geometry and charge distribution but different structural topology, indicating convergent evolution of toxin domains for optimized host target interaction.
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Magnesium chloride, ≥98%
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
1,3-Propanediol, 98%
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Magnesium chloride, suitable for insect cell culture, BioReagent, ≥97.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~0.025 M in H2O
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Sigma-Aldrich
Magnesium chloride solution, Molecular Biology, 1.00 M±0.01 M
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, 2 M in H2O