Merck
  • Study of the Biotransformation of Tongmai Formula by Human Intestinal Flora and Its Intestinal Permeability across the Caco-2 Cell Monolayer.
Study of the Biotransformation of Tongmai Formula by Human Intestinal Flora and Its Intestinal Permeability across the Caco-2 Cell Monolayer.
Molecules (Basel, Switzerland) (2015-10-27)
Shuai Wu, Wei Xu, Fu-Rong Wang, Xiu-Wei Yang
ABSTRACT
Tongmai formula (TMF) is a well-known Chinese medicinal preparation that contains isoflavones as its major bioactive constituents. As traditional Chinese medicines (TCMs) are usually used by oral administration, their fate inside the intestinal lumen, including their biotransformation by human intestinal flora (HIF) and intestinal absorption deserves study. In this work TMF extract was incubated with human intestinal bacteria under anaerobic conditions and the changes in the twelve main constituents of TMF were then investigated. Their intestinal permeabilities, i.e., the transport capability across the intestinal brush border were investigated with a human colon carcinoma cell line (Caco-2) cell monolayer model to predict the absorption mechanism. Meanwhile, rapid HPLC-DAD methods were established for the assay. According to the biotransformation curves of the twelve constituents and the permeability coefficients, the intestinal absorption capacity of the typical compounds was elevated from the levels of 10(-7) cm/s to 10(-5) cm/s from those of the original compounds in TMF. Among them the main isoflavone glycosides puerarin (4), mirificin (6) and daidzin (7) were transformed into the same aglycone, daidzein (10). Therefore it was predicted that the aglycone compounds might be the real active ingredients in TMF. The models used can represent a novel path for the TCM studies.
Sigma-Aldrich
MES solution, BioUltra, Molecular Biology, 0.5 M in H2O
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Naringin, ≥95% (HPLC)
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Naringin, ≥90% (HPLC), from citrus fruit
Sigma-Aldrich
MES, low moisture content, ≥99% (titration)
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
L-Cysteine hydrochloride, anhydrous, ≥99.0% (RT)
Sigma-Aldrich
Sodium thioglycolate, ≥96.5% (iodometric)
Sigma-Aldrich
L-Cysteine hydrochloride, anhydrous, ≥98% (TLC)
Sigma-Aldrich
Atenolol, ≥98% (TLC), powder
Sigma-Aldrich
L-Cysteine hydrochloride, anhydrous, from non-animal source, BioReagent, suitable for cell culture, ≥98.0%