Merck
  • Bis(1,2-dithiosquarato)nickelates(II) - Synthesis, Structure, EPR and Thermal Behavior.
Bis(1,2-dithiosquarato)nickelates(II) - Synthesis, Structure, EPR and Thermal Behavior.
Acta chimica Slovenica (2015-06-19)
Peter Strauch, Mike Neumann, Alexandra Kelling, Uwe Schilde
ABSTRACT
1,2-Dithiosquaratonickelates are available by direct synthesis from metal salts with dipotassium-1,2-dithiosquarate and the appropriate counter cations. The synthesis and characterization, including mass spectrometry, of a series 1,2-dithiosquaratonickelates(II), [Ni(dtsq)2]2-, with several "onium" cations is reported and the X-ray structures of two diamagnetic complexes, (HexPh3N)2[Ni(dtsq)2] and (BuPh3N2[Ni(dtsq)2] with sterically demanding counter ions are presented. The diamagnetic nickel complexes have been doped as host lattices with traces of Cu(II) to measure EPR for additional structural information. The thermal behavior of this series is studied by thermogravimetry and differential thermo analysis (TG/DTA). The thermolysis in air as well as under nitrogen atmosphere of these complexes results in nickel oxide nano-particles in all cases, which are characterized by X-ray powder diffraction.
Sigma-Aldrich
Potassium bromide, anhydrous, powder, 99.95% trace metals basis
Sigma-Aldrich
Potassium bromide, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
Potassium bromide, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Diisopropyl ether, anhydrous, 99%, contains either BHT or hydroquinone as stabilizer
Sigma-Aldrich
Potassium bromide, BioXtra, ≥99.0%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis