Merck
  • Large work function difference driven electron transfer from electrides to single-walled carbon nanotubes.
Large work function difference driven electron transfer from electrides to single-walled carbon nanotubes.
Nanoscale (2014-06-24)
Mini Mol Menamparambath, Jong-Ho Park, Ho-Sung Yoo, Shashikant P Patole, Ji-Beom Yoo, Sung Wng Kim, Seunghyun Baik
ABSTRACT
A difference in work function plays a key role in charge transfer between two materials. Inorganic electrides provide a unique opportunity for electron transfer since interstitial anionic electrons result in a very low work function of 2.4-2.6 eV. Here we investigated charge transfer between two different types of electrides, [Ca(2)N](+)·e(-) and [Ca(24)Al(28)O(64)](4+)·4e(-), and single-walled carbon nanotubes (SWNTs) with a work function of 4.73-5.05 eV. [Ca(2)N](+) · e(-) with open 2-dimensional electron layers was more effective in donating electrons to SWNTs than closed cage structured [Ca(24)Al(28)O(64)](4+) · 4e(-) due to the higher electron concentration (1.3 × 10(22) cm(-3)) and mobility (∼ 200 cm(2) V(-1) s(-1) at RT). A non-covalent conjugation enhanced near-infrared fluorescence of SWNTs as high as 52%. The field emission current density of electride-SWNT-silver paste dramatically increased by a factor of 46,000 (14.8 mA cm(-2)) at 2 V μm(-1) (3.5 wt% [Ca(2)N](+) · e(-)) with a turn-on voltage of 0.85 V μm(-1).
Sigma-Aldrich
4-Methyl-2-pentanone, suitable for HPLC, ≥99.5%
Pricing and availability is not currently available.