Merck
  • Iron casein succinylate-chitosan coacervate for the liquid oral delivery of iron with bioavailability and stability enhancement.
Iron casein succinylate-chitosan coacervate for the liquid oral delivery of iron with bioavailability and stability enhancement.
Archives of pharmacal research (2015-12-01)
Kyoung Ah Min, Jung-Hye Cho, Yun-Kyoung Song, Chong-Kook Kim
ABSTRACT
Iron casein succinylate (ICS) liquid oral preparation as iron supplement has uncomfortable taste after a long period of storage because of its stability, and poor bioavailability of iron compared to any other iron preparations. To improve the chemical stability of ICS and enhance the bioavailability of iron, chitosan-ICS nanoparticles (NPs) were prepared by complex coacervation method and stabilized with polyethylene glycol (PEG) 400. NPs were spherical (mean diameter of 830-1070 nm) with positive charge (+30-60 mV) depending on the composition of NPs. Addition of PEG400 (2 w/v %) increased the zeta potential (26-50 %) and physical stability of chitosan-ICS NPs suspension. Also, NPs decreased iron release compared to ICS after 7-weeks of storage at 4 °C. NPs markedly increased the permeability of iron in Caco-2 cell up to 32-38-fold compared to ICS, while physical mixture of chitosan and ICS increased the iron permeability only 2.5-fold. In summary, NPs improved the physicochemical stability and enhanced the transport of iron compared to other iron preparations in Caco-2 cell model. Thus, chitosan-ICS coacervate might be a promising candidate as a liquid oral iron delivery system for iron deficiency patients with stability and bioavailability enhancement.
Sigma-Aldrich
Pepsin from porcine gastric mucosa, powder, ≥250 units/mg solid
Sigma-Aldrich
Pancreatin from porcine pancreas, 4 × USP specifications