- Novel solvent properties of choline chloride/urea mixtures.
Novel solvent properties of choline chloride/urea mixtures.
Chemical communications (Cambridge, England) (2003-03-04)
Andrew P Abbott, Glen Capper, David L Davies, Raymond K Rasheed, Vasuki Tambyrajah
PMID12610970
ABSTRACT
Eutectic mixtures of urea and a range of quaternary ammonium salts are liquid at ambient temperatures and have interesting solvent properties.