Merck
  • Sensitive, nonradioactive assay of phosphorylase kinase through measurement of enhanced phosphorylase activity towards fluorogenic dextrin.
Sensitive, nonradioactive assay of phosphorylase kinase through measurement of enhanced phosphorylase activity towards fluorogenic dextrin.
Journal of biochemistry (2015-09-18)
Daichi Miyagawa, Yasushi Makino, Masaaki Sato
ABSTRACT
Glycogen phosphorylase (GP) exists in two interconvertible forms, GPa (phosphorylated form, high activity) and GPb (nonphosphorylated form, low activity). Phosphorylase kinase (PhK) catalyses the phosphorylation of GPb and plays a key role in the cascade system for regulating glycogen metabolism. In this study, we developed a highly sensitive and nonradioactive assay for PhK activity by measuring the enhanced GP activity towards a pyridylaminated maltohexaose. The enhanced GP activity (ΔA) was calculated by the following formula: ΔA = A(+) - A(0), where A(+) and A(0) represent the GP activities of the PhK-treated and PhK-nontreated samples, respectively. Using a high-performance liquid chromatograph equipped with a fluorescence spectrophotometer, the product of GP activity could be isolated and quantified at 10 fmol. This method does not require the use of any radioactive compounds and only 1 µg of GPb per sample was needed to obtain A(+) and A(0) values. The remarkable reduction in GPb concentration enabled us to discuss an interesting new role for glycogen in PhK activity.
Sigma-Aldrich
Maltohexaose, ≥65% (HPLC)