Merck
  • ERK3 regulates TDP2-mediated DNA damage response and chemoresistance in lung cancer cells.
ERK3 regulates TDP2-mediated DNA damage response and chemoresistance in lung cancer cells.
Oncotarget (2015-12-25)
Ka Bian, Naveen Reddy Muppani, Lobna Elkhadragy, Wei Wang, Cheng Zhang, Tenghui Chen, Sungyun Jung, Ole Morten Seternes, Weiwen Long
ABSTRACT
Posttranslational modifications (PTMs), such as phosphorylation and ubiquitination, play critical regulatory roles in the assembly of DNA damage response proteins on the DNA damage site and their activities in DNA damage repair. Tyrosyl DNA phosphodiesterase 2 (TDP2) repairs Topoisomerase 2 (Top2)-linked DNA damage, thereby protecting cancer cells against Top2 inhibitors-induced growth inhibition and cell death. The regulation of TDP2 activity by post-translational modifications in DNA repair, however, remains unclear. In the current study, we have found that ERK3, an atypical MAPK, phosphorylates TDP2 at S60 and regulates TDP2's phosphodiesterase activity, thereby cooperatively protecting lung cancer cells against Top2 inhibitors-induced DNA damage and growth inhibition. As such, our study revealed a post-translational regulation of TDP2 activity and discovered a new role of ERK3 in increasing cancer cells' DNA damage response and chemoresistance to Top2 inhibitors.
Sigma-Aldrich
Phosphatase Inhibitor Cocktail 3, DMSO solution
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Ttrap
Sigma-Aldrich
Etoposide, synthetic, 95.0-105.0%, powder