Merck
  • Studies on insecticidal activities and action mechanism of novel benzoylphenylurea candidate NK-17.
Studies on insecticidal activities and action mechanism of novel benzoylphenylurea candidate NK-17.
PloS one (2013-06-19)
Yongqiang Li, Yaoguo Qin, Na Yang, Yufeng Sun, Xinling Yang, Ranfeng Sun, Qingmin Wang, Yun Ling
ABSTRACT
Insecticidal activity of NK-17 was evaluated both in laboratory and in field. It was found that the toxicity of NK-17 against S. exigua was 1.93 times and 2.69 times those of hexaflumuron and chlorfluazuron respectively, and the toxicity of NK-17 against P. xylostella was 1.36 times and 1.90 times those of hexaflumuron and chlorfluazuron respectively, and the toxicity of NK-17 against M. separate was 18.24 times those of hexaflumuron in laboratory, and 5% NK-17 EC at 60 g a.i ha(-1) can control S. exigua and P. xylostella with the best control efficiency of about 89% and over 88% respectively in Changsha and Tianjin in field. The insecticidal mechanism of NK-17 was explored for the first time by utilizing the fluorescence polarization method. NK-17 could bind to sulfonylurea receptor (SUR) of B. germanica with stronger affinity comparing to diflubenzuron and glibenclamide, which suggested that NK-17 may also act on the site of SUR to inhibit the chitin synthesis in insect body and the result can well explain that NK-17 exhibited stronger toxicity against B. germanica than diflubenzuron and glibenclamide in vivo.
Product Description
Supelco
Hexaflumuron, PESTANAL®, analytical standard
Supelco
Diflubenzuron, PESTANAL®, analytical standard